Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200618

Sigma-Aldrich

Monoclonal Anti-AXL antibody produced in mouse

~1.0 mg/mL, clone AXL188, purified immunoglobulin

Synonym(s):

Anti-AXL oncogene, Anti-AXL receptor tyrosine kinase, Anti-AXL transforming sequence/gene, Anti-JTK11, UFO, Anti-tyrosine-protein kinase receptor UFO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AXL188, monoclonal

form

buffered aqueous solution

mol wt

antigen ~130 kDa

species reactivity

human, monkey

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 20 μg/test using A549 cells
immunoblotting: 0.5-1.0 μg/mL using A549 total cell extracts
immunofluorescence: 20 μg/mL using A549 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AXL(558)

General description

Monoclonal Anti-AXL (mouse IgG1 isotype) is derived from the hybridoma AXL188 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to an internal sequence of human AXL. AXL (Adhesion-related kinase) is a part of TAM (Tyro3, AXL and Mertk) family of receptor tyrosine kinases. Its ligand is Gas6 (Growth arrest specific gene 6). It is located on the human chromosome at 19q13.2.

Immunogen

synthetic peptide corresponding to an internal sequence of human AXL

Application

Monoclonal Anti-AXL antibody has been used in
  • immunoblotting
  • immunocytochemistry
  • immunofluorescence
  • flow cytometry

Biochem/physiol Actions

Activated AXL (Adhesion-related kinase) elicits a protective action in cells from apoptosis. It also increases migration, aggregation and growth by activating several downstream pathways. AXL has been found to play a significant role in pathogenesis and progression of human cancers including breast, hepatic cellular carcinoma, thyroid, esophageal, astrocytoma-glioblastoma. It was also suggested to serve as an important biomarker for disease status and prognosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting Axl and Mer kinases in cancer
Verma A, et al.
Molecular Cancer Therapeutics, 10(10), 1763-1773 (2011)
Axl-dependent signalling: a clinical update
Korshunov V A.
Clinical Science, 122(8), 361-368 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service