Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200607

Sigma-Aldrich

Anti-Claudin-3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Cldn3, Rat ventral prostate 1 protein (RVP1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

buffered aqueous solution

species reactivity

human, rat

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using A549 cells extracts
immunohistochemistry: 5-10 μg/mL using formalin-fixed, paraffin-embedded rat intestine/duodenum sections

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN3(1365)
rat ... Cldn3(65130)

General description

Claudin-3 (CLDN3) is located on the human chromosome at 7q11.23.

Specificity

Anti-Claudin-3 recognizes human and rat Claudin-3.

Immunogen

a synthetic peptide corresponding to an internal sequence of rat Caludin-3, conjugated to KLH

Application

Anti-Claudin-3 antibody produced in rabbit may be used in:
  • immunoblotting
  • immunohistochemistry
  • immunofluorescence

Biochem/physiol Actions

Claudin-3 (CLDN3) promotes interactions with other cells in vivo that prevent growth and metastatic potential by assisting the expression of E-cadherin and limiting β-catenin signaling. CLDN3 represents a promising target for cancer detection, diagnosis and therapy. Overexpression of CLDN3 is associated with several neoplasias, such as ovarian, breast, prostate and pancreatic cancers. CLDN3 levels are increased in lung adenocarcinoma cells.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tight junction proteins claudin-3 and claudin-4 control tumor growth and metastases
Shang X, et al.
Neoplasia, 14(10), 974-IN22-974-IN22 (2012)
Williams syndrome deficits in visual spatial processing linked to GTF2IRD1 and GTF2I on chromosome 7q11. 23
Hirota H, et al.
Genetics in Medicine : Official Journal of the American College of Medical Genetics, 5(4), 311-321 (2003)
Claudin-3 expression increases the malignant potential of lung adenocarcinoma cells: role of epidermal growth factor receptor activation
Zhang L, et al.
Testing, 8(14), 23033-23033 (2017)
Claudin proteins in human cancer: promising new targets for diagnosis and therapy
Morin P J
Cancer Research, 65(21), 9603-9606 (2005)
Probiotics Prevents Sensitization to Oral Antigen and Subsequent Increases in Intestinal Tight Junction Permeability in Juvenile-Young Adult Rats
Tulyeu J, et al.
Microorganisms, 7(10), 463-463 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service