Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200596

Sigma-Aldrich

Anti-NSF (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-N-ethylmaleimide-sensitive factor-like protein, Anti-N-ethylmaleimide-sensitive fusion protein, Anti-NEM-sensitive fusion protein, Anti-vesicle-fusing ATPase, Anti-vesicular-fusion protein NSF, SKD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using extracts of SH-SY5Y cells.
immunohistochemistry: 10-20 μg/mL using formalin-fixed, paraffin-embedded rat brain.
immunoprecipitation (IP): 10 μg using lysates of mouse brain (S1 fraction).
indirect immunofluorescence: 1-2 μg/mL using Neuro-2A cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NSF(4905)

General description

N-ethylmaleimide-sensitive fusion protein (NSF) is a homotrimer and is made up of three domains such as N-terminal domain and two homologous adenosine triphosphate (ATP)-binding domains. It is located on the human chromosome at 17q21.31.

Specificity

Anti-NSF(C-terminal) specifically recognizes human, rat and mouse NSF.

Immunogen

synthetic peptide corresponding to a sequence located in the C-terminus of human NSF, conjugated to KLH. The corresponding sequence is highly conserved (single amino acid substitution) in rat and mouse NSF.

Application

Anti-NSF (C-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry
  • immunohistochemistry
  • immunofluorescence

Biochem/physiol Actions

N-ethylmaleimide-sensitive fusion protein (NSF) is an essential component of the protein machinery responsible for various membrane fusion events, including inter-cisternal Golgi protein transport and the exocytosis of vesicles. NSF takes part in NSF-dependent membrane fusion in which interaction of two types of cytosolic proteins, NSF and α-, β- and γ- soluble NSF attachment protein (SNAP) isoforms, with the subcellular compartment-specific SNAP receptors (SNAREs) of the vesicle and target membranes. NSF is required in long-term potentiation (LTP), underlying the formation of long-term memory, by regulating the exocytosis of glutamate α-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid (AMPA) glutamate receptor 2 (GluR2) at postsynaptic densities (PSD).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use,store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction between N-ethylmaleimide-sensitive factor and GluR2 is essential for fear memory formation in lateral amygdala
Joels G and Lamprecht R
The Journal of Neuroscience, 30(47), 15981-15986 (2010)
N-ethylmaleimide-sensitive fusion protein: a trimeric ATPase whose hydrolysis of ATP is required for membrane fusion.
Whiteheart S W, et al.
The Journal of Cell Biology, 126(4), 945-954 (1994)
Cellular functions of NSF: not just SNAPs and SNAREs
Zhao C, et al.
Febs Letters, 581(11), 2140-2149 (2007)
The AMPA receptor GluR2 C terminus can mediate a reversible, ATP-dependent interaction with NSF and alpha-and beta-SNAPs
Osten P, et al.
Neuron, 21(1), 99-110 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service