Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200587

Sigma-Aldrich

Anti-CHD1 antibody from rabbit

affinity isolated antibody

Synonym(s):

ATP-dependent helicase CHD1, chromodomain helicase DNA binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

buffered aqueous solution

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 5-10 μg/mL using whole extracts of mouse P19 and human HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHD1(1105)
mouse ... Chd1(12648)

General description

Chromodomain-helicase-DNA-binding protein 1 (CHD1) is an adenosine triphosphate (ATP)-dependent chromatin-remodeling factor. It belongs to the CHD family of proteins. CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD1 gene is mapped on the human chromosome at 5q15-q21.1.

Specificity

Anti- CHD1 recognizes human and mouse CHD1.

Immunogen

synthetic peptide corresponding to an internal region of human CHD1 , conjugated to KLH.

Application

Anti-CHD1 antibody from rabbit may be used in immunoblotting.

Biochem/physiol Actions

Chromodomain-helicase-DNA-binding protein 1 (CHD1) is required for maintaining open chromatin and pluripotency in embryonic stem cells. It is involved in gametogenesis. CHD1 plays a role in the incorporation of H3.3 into male pronucleus during the process of decondensation post-fertilization.

Physical form

a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromosomal abnormalities and novel disease-related regions in progression from Barrett's esophagus to esophageal adenocarcinoma
Akagi T, et al.
International Journal of Cancer. Journal International Du Cancer, 125(10), 2349-2359 (2009)
Lena Ho et al.
Nature, 463(7280), 474-484 (2010-01-30)
New methods for the genome-wide analysis of chromatin are providing insight into its roles in development and their underlying mechanisms. Current studies indicate that chromatin is dynamic, with its structure and its histone modifications undergoing global changes during transitions in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service