Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200584

Sigma-Aldrich

Anti-COG2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Component of oligomeric Golgi complex 2, Anti-LDLC, Anti-Low density lipoprotein receptor defect C-complementing protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~83 kDa

species reactivity

human

technique(s)

immunoblotting: 1:1000-1:2000 using whole extracts of hamster CHO cells.
immunoprecipitation (IP): 2.5-5.0 μL using lysates of human HeLa cells
indirect immunofluorescence: 1:2,500-1:5,000 using human HeLa cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG2(22796)

General description

Conserved oligomeric Golgi (COG) complex 2 is also known as LDLC. COG complex is a multi-subunit protein complex, composed of eight distinct subunits. This protein is highly conserved with two heterotrimeric groups, Cog2-Cog3- Cog4 and Cog5-Cog6-Cog7 in its structure.

Immunogen

synthetic peptide corresponding to the C-terminal region of human COG2, conjugated to KLH. The corresponding sequence is identical in monkey and bovine and differs by four amino acids in rat and mouse COG2.

Application

Anti-COG2 antibody produced in rabbit has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Conserved oligomeric Golgi (COG) complex 2 regulates membrane trafficking and maintenance of Golgi glycosylation machinery in eukaryotic cells. COG2 is needed for normal Golgi morphology and function. COG2 interacts with p115 (golgin), a vesicle tethering protein that functions in endoplasmic reticulum-Golgi trafficking. This interaction is required for Golgi integrity.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The interaction of two tethering factors, p115 and COG complex, is required for Golgi integrity
Sohda M, et al.
Traffic, 8(3), 270-284 (2007)
Comparative analyses of the Conserved Oligomeric Golgi (COG) complex in vertebrates
Quental R, et al.
BMC Evolutionary Biology, 10(1), 212-212 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service