Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200583

Sigma-Aldrich

Anti-Epsin1 (internal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90 kDa

species reactivity

rat, mouse, human

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using extracts of SH-SY5Y cells and of mouse brain (S1 fraction).
indirect immunofluorescence: 2-4 μg/mL using NRK cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPN1(29924)
mouse ... Epn1(13854)
rat ... Epn1(117277)

General description

Epsin1 is an endocytic adaptor protein. It contains an epsin N-terminal homology (ENTH) region, a single clathrin-binding LVDLD motif and two or three tandem ubiquitin-interacting motifs (UIMs). It is localized on the human chromosome at 19q13.42.

Specificity

Anti-Epsin1 (internal) specifically recognizes human,rat and mouse Epsin1.

Immunogen

synthetic peptide corresponding to an internal sequence of human epsin1 isoform A, conjugated to KLH. The corresponding sequence is identical in human espin1 isoforms B and C, and highly conserved (82% identity) in rat and mouse epsin1.

Application

Anti-Epsin1 (internal) antibody produced in rabbit may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Epsin1 plays a central role in the clathrin-mediated internalization and degradation of ubiquitinated proteins. It interacts with Eps15 subunit of the clathrin adaptor AP2, clathrin and other accessory proteins. Epsin1 plays a role in the recruitment of ubiquitinated epidermal growth factor receptor (EGFR) into clathrin-coated pits. Epsin1 is required in the mitotic membrane and spindle organization. Overexpression of epsin1 is associated with fibrosarcoma cell invasion.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epsin: inducing membrane curvature
Horvath C A J, et al.
The International Journal of Biochemistry & Cell Biology, 39(10), 1765-1770 (2007)
A genotype resource for postmortem brain samples from the Autism Tissue Program
Wintle R F, et al.
Autism Research : Official Journal of the International Society for Autism Research, 4(2), 89-97 (2011)
The epsin family of endocytic adaptors promotes fibrosarcoma migration and invasion
Coon BG, et al.
The Journal of Biological Chemistry, 285(43), 33073-33081 (2010)
Epsin 1 is involved in recruitment of ubiquitinated EGF receptors into clathrin-coated pits
Kazazic M, et al.
Traffic, 10(2), 235-245 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service