Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200573

Sigma-Aldrich

Anti-Claudin-4 antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CLDN4, Anti-CPE-R, Anti-CPER, Anti-CPETR, Anti-CPETR1, Anti-WBSCR8, Anti-hCPE-R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~19 kDa

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 1.5-3.0 μg/mL using Extracts of HEK-293T cells overexpressing human claudin-4.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN4(1364)

General description

Claudin-4 is heterogeneously expressed in various tissues. It is enriched in endothelial cells (ECs), including the blood-brain barrier, and is also expressed in liver, gut and kidney. Claudin-4 is a major constituent of tight junctions.

Specificity

Anti-Claudin-4 specifically recognizes humanclaudin-4.

Immunogen

synthetic peptide corresponding to an internal sequence of human claudin-4, conjugated to KLH. The corresponding sequence is highly conserved in mouse claudin-4 (89% identity) and in rat claudin-4 (84% identity).

Application

Anti-Claudin-4 antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Claudin-4 expression is deregulated in several cancers, with aberrant expression in ovarian, breast, prostate, gastric and pancreatic carcinomas.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel tumor-targeted therapy using a claudin-4-targeting molecule
Saeki R, et al.
Molecular Pharmacology, 76(4), 918-926 (2009)
Adele Moatti et al.
STAR protocols, 4(2), 102220-102220 (2023-04-16)
The inner ear of humans and large animals is embedded in a thick and dense bone that makes dissection challenging. Here, we present a protocol that enables three-dimensional (3D) characterization of intact inner ears from large-animal models. We describe steps
Claudin-3 and claudin-4 expression in ovarian epithelial cells enhances invasion and is associated with increased matrix metalloproteinase-2 activity
Agarwal R, et al.
Cancer Research, 65(16), 7378-7385 (2005)
Claudin-4 expression decreases invasiveness and metastatic potential of pancreatic cancer
Michl P, et al.
Cancer Research, 63(19), 6265-6271 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service