Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200556

Sigma-Aldrich

Anti-BMP4 antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-BMP2B, Anti-BMP2B1, Anti-Bone morphogenetic protein 4, ZYME, Anti-MCOPS6, Anti-OFC11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 20 μg/mL using formalin-fixed, paraffin-embedded human colon.
immunoprecipitation (IP): 10 μg using lysates of CHO cells overexpressing human BMP4.
western blot: 1-2 μg/mL using extracts of CHO cells overexpressing human BMP4.

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP4(652)

General description

Bone morphogenetic proteins (BMPs) are members of the TGF-β superfamily. BMP4, also known as ZYME or BMP2B, is encoded by the gene mapped to human chromosome 14q22.2. BMPs are expressed in localized patterns during embryonic mouse brain development.

Specificity

Anti-BMP4 specifically recognizes human BMP4.

Immunogen

synthetic peptide corresponding to an internal sequence of human BMP4, conjugated to KLH. The corresponding sequence is highly conserved (single amino acid substitution) in rat and mouse BMP4.

Application

Anti-BMP4 antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunohistochemistry

Biochem/physiol Actions

Bone morphogenetic proteins (BMPs) were originally identified as growth factors that can induce bone and cartilage formation. Various members of the BMP family are involved in a large variety of functions during development and in cancer. BMP4 is implicated in regulation of cell proliferation, differentiation, apoptosis, adult tissue homeostasis and cell fate determination during development. Specifically, BMP4 has been shown to regulate mesodermal cell commitment to the hematopoietic cell lineage during early embryogenesis. BMP4 is crucial for development since knockout mice die early in embryogenesis. Overexpression of BMP4 has been reported in several cancers, including melanoma, hepatocellular carcinoma, adenocarcinoma and colorectal cancer and is frequently accompanied by an enhanced cell migratory and invasive potential of the tumors.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bone morphogenetic protein-4 is overexpressed in colonic adenocarcinomas and promotes migration and invasion of HCT116 cells
Deng H, et al.
Experimental Cell Research, 313(5), 1033-1044 (2007)
Bone Morphogenetic Protein (BMP) signaling in development and human diseases
Wang RN, et al.
Genes & diseases, 1(1), 87-105 (2014)
The 14q22. 2 colorectal cancer variant rs4444235 shows cis-acting regulation of BMP4
Lubbe SJ, et al.
Oncogene, 31(33), 3777-3784 (2012)
BMP4 regulates the hematopoietic stem cell niche
Goldman D C, et al.
Blood, 114(20), 4393-4401 (2009)
Bone morphogenetic protein 4 is induced in hepatocellular carcinoma by hypoxia and promotes tumour progression
Maegdefrau U, et al.
Journal of pathogens, 218(4), 520-529 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service