Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200546

Sigma-Aldrich

Anti-SLAMF6 antibody, Mouse monoclonal

clone SF24, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-SLAMF6 antibody produced in mouse, Monoclonal Anti-Activating NK receptor, Monoclonal Anti-CD352, Monoclonal Anti-KALI NTB-A, Monoclonal Anti-KALIb, Monoclonal Anti-Ly108, Monoclonal Anti-NK-T-B-antigen, Monoclonal Anti-NTBA, Monoclonal Anti-NTBA receptor, Monoclonal Anti-SF2000, Monoclonal Anti-SLAM family member 6, Monoclonal Anti-natural killer-, T- and B-cell antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SF24, monoclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 10-20 μg/test using MCF7 cells.
indirect immunofluorescence: 10-20 μg/mL using MCF7 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLAMF6(114836)

General description

Monoclonal Anti-SLAMF6 (mouse IgM isotype) is derived from the hybridoma SF24 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Signaling lymphocyte activation molecule family member 6 (SLAMF6) belongs to CD2 receptor subfamily of immunoglobulin (Ig) superfamily. It is a type I transmembrane glycoprotein. SLAMF6 consists of two extracellular immunoglobulin (Ig)-like domains and three cytoplasmic tyrosine-based signaling motifs. It is present on the surface of immune cells such as B-cells, T-cells and natural killer (NK) cells.

Specificity

Monoclonal Anti-SLAMF6 recognizes human SLAMF6.

Immunogen

synthetic peptide corresponding to an internal sequence of human SLAMF6. The isotype is determined by ELISA using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-SLAMF6 antibody produced in mouse may be used in:
  • immunoblotting
  • flow cytometry
  • immunocytochemistry

Biochem/physiol Actions

Signaling lymphocyte activation molecule family member 6 (SLAMF6) takes part in the uncoupling of SLAM receptors from Src homology 2 domain-containing protein tyrosine phosphatase 1 (SHP-1). Overexpression of SLAMF6 is observed in B-lymphocytes from chronic lymphocytic leukemia (CLL) and lymphoma patients.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gntb-A, a Novel Sh2d1a-Associated Surface Molecule Contributing to the Inability of Natural Killer Cells to Kill Epstein-Barr Virus-Infected B Cells in X-Linked Lymphoproliferative Disease
Bottino C, et al.
The Journal of Experimental Medicine, 194(3), 235-246 (2001)
Slamf6 negatively regulates autoimmunity
Wang N, et al.
Clinical Immunology (Orlando, Fla.), 173(4), 19-26 (2016)
A hematopoietic cell-driven mechanism involving SLAMF6 receptor, SAP adaptors and SHP-1 phosphatase regulates NK cell education
Wu N, et al.
Nature Immunology, 17(4), 387-396 (2016)
SLAMF6 in health and disease: Implications for therapeutic targeting
Yigit B, et al.
Clinical Immunology (Orlando, Fla.) (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service