Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200525

Sigma-Aldrich

Anti-Kindlin-2 antibody, Mouse monoclonal

clone 3A3, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-KIND2, Monoclonal Anti-Kindlin-2 antibody produced in mouse, Monoclonal Anti-MIG2, Monoclonal Anti-Mitogen-inducible gene 2 protein, Monoclonal Anti-PH domain-containing family C member 1, Monoclonal Anti-PLEKHC1, Monoclonal Anti-UNC112, Monoclonal Anti-UNC112B, Monoclonal Anti-family C (with FERM domain) member 1, Monoclonal Anti-fermitin family member 2, Monoclonal Anti-mig-2, Monoclonal Anti-mitogen inducible gene 2 protein, Monoclonal Anti-pleckstrin homology domain containing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

3A3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

bovine, monkey, human, mouse, rat, dog

concentration

~1.0 mg/mL

technique(s)

western blot: 1.0-2.0 μg/mL using HeLa total cell extracts

isotype

IgG1

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-Kindlin-2 (mouse IgG1 isotype) is derived from the hybridoma 3A3 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Kindlin-2 belongs to the Kindlin family of focal adhesion proteins. It is localized in all solid tissues of mesenchymal origin. Kindlin-2 consists of band 4.1 protein, ezrin, radixin and moesin (FERM) and pleckstrin homology (PH) domains. It is mapped on the human chromosome at 14q22.1.

Specificity

Monoclonal Anti-Kindlin-2 recognizes human, monkey, bovine, dog rat and mouse kindlin-2.

Immunogen

recombinant C-terminal region fusion protein of human Kindlin-2.1 The isotype is determined by ELISA using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-Kindlin-2 antibody produced in mouse may be used in:
  • immunoblotting
  • flow cytometry
  • immunoprecipitation
  • immunocytochemistry
  • immunohistochemistry

Biochem/physiol Actions

Kindlin-2 plays an important role in integrin-mediated cell-extracellular matrix (ECM) adhesion and invasion. It is also involved in wound healing, angiogenesis and embryonic development. Deficiency of kindlin-2 leads to the lethality of embryos. Kindlin-2 expression can be altered by malignant transformations such as breast cancer and leiomyomas.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kindlin-2 controls bidirectional signaling of integrins
Montanez, et al.
Genes & Development, 22(10), 1325-1330 (2008)
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus
Moretti F, et al.
eLife, 7(10), e35816-e35816 (2018)
Kindlin-2 regulates podocyte adhesion and fibronectin matrix deposition through interactions with phosphoinositides and integrins
Qu H, et al.
Journal of Cell Science, 124(6), 879-891 (2011)
Kindlin-2: a novel prognostic biomarker for patients with hepatocellular carcinoma
Ge Y, et al.
Pathology Research and Practice, 211(3), 198-202 (2015)
Kindlins in FERM adhesion
Malinin NL, et al.
Blood, 115(20), 4011-4017 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service