Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200516

Sigma-Aldrich

Anti-BMP4 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-BMP2B, Anti-BMP2B1, Anti-Bone morphogenetic protein 4, ZYME, Anti-MCOPS6, Anti-OFC11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 20 μg/mL using formalin-fixed, paraffin-embedded human colon.
western blot: 1.0-2.0 μg/mL using extracts of CHO cells overexpressing human BMP4.

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP4(652)
mouse ... Bmp4(12159)
rat ... Bmp4(25296)

General description

Bone morphogenetic protein 4 (BMP4) protein is a member of the transforming growth factor-β (TGF-β) superfamily. BMP4 gene is located at 14q22.2 on the human chromosome.

Specificity

Anti-BMP4 (N-terminal) specifically recognizes human BMP4.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal region of human BMP4, conjugated to KLH. The corresponding sequence is identical in rat and mouse BMP4.

Application

Anti-BMP4 (N-terminal) antibody produced in rabbit may be used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Bone morphogenetic protein 4 (BMP4) protein is involved in the regulation of cell proliferation and differentiation. It is also involved in apoptosis and determination of cell fate during development. BMP4 plays a role in the regulation of mesodermal cell commitment to the hematopoietic cell lineage during early embryogenesis. Overexpression of BMP4 gene has been reported in several cancers, including melanoma, hepatocellular carcinoma, adenocarcinoma and colorectal cancer.It is frequently accompanied by an enhanced cell migratory and invasive potential of the tumors.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BMP4 regulates the hematopoietic stem cell niche
Goldman D C, et al.
Blood, 114(20), 4393-4401 (2009)
Multiple common susceptibility variants near BMP pathway loci GREM1, BMP4, and BMP2 explain part of the missing heritability of colorectal cancer
Tomlinson IPM, et al.
PLoS Genetics, 7(6), e1002105-e1002105 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service