Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200512

Sigma-Aldrich

Anti-KIAA1033 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-WASH complex subunit 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~130 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

western blot: 3-6 μg/mL using whole extracts of HEK-293T cells overexpressing human KIAA1033

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

synthetic peptide corresponding to an internal sequence of human KIAA1033, conjugated to KLH. The corresponding sequence is identical in monkey, bovine and pig and differs by one amino acid in mouse and rat.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service