Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200507

Sigma-Aldrich

Anti-ATG101 antibody, Mouse monoclonal

clone ATG101-7, purified from hybridoma cell culture

Synonym(s):

Anti-C12orf44, chromosome 12 open reading frame 44, PP894, Atg13-interacting protein, Anti-autophagy-related protein 101, Monoclonal Anti-ATG101 antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ATG101-7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extracts of HEK-293T cells overexpressing human ATG101.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-ATG101 (mouse IgG1 isotype) is derived from the hybridoma ATG101-7 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Atg101 is conserved in humans and several eukaryotes. It is not conserved in yeast Saccharomyces cerevisiae. ATG101 gene is located on the human chromosome at 12q13.13.

Specificity

Anti- ATG101 recognizes human ATG101.

Immunogen

synthetic peptide corresponding to the N-terminal region of human ATG101, conjugated to KLH. The corresponding sequence is identical in mouse, rat, monkey, canine and bovine ATG101.

Application

Monoclonal Anti-ATG101 antibody produced in mouse is suitable for use in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Atg101 is a mammalian Atg13 binding protein essential for autophagagy. Atg13 forms a stable complex with Unc-51 like autophagy activating kinase 1 (ULK1) and focal adhesion kinase (FAK) family kinase-interacting protein of 200 kDa (FIP200). Atg101 is linked to the ULK1-Atg13-FIP200 complex through direct contact with Atg13.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20°C in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of Androgen Responsive Elements in Some Autophagy Related Genes via In Silico Analysis.
Erzurumlu Y
Journal of Health Science, 11(1) (2020)
A novel, human Atg13 binding protein, Atg101, interacts with ULK1 and is essential for macroautophagy
Mercer C A, et al.
Autophagy, 5(5), 649-662 (2009)
Structure of the human Atg13-Atg101 HORMA heterodimer: an interaction hub within the ULK1 complex
Qi S, et al.
Structure, 23(10), 1848-1857 (2015)
Atg101, a novel mammalian autophagy protein interacting with Atg13
Hosokawa N, et al.
Autophagy, 5(7), 973-979 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service