Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200505

Sigma-Aldrich

Anti-DIS3 antibody, Mouse monoclonal

clone DIS3-N, purified from hybridoma cell culture

Synonym(s):

Anti-EXOSC11, Anti-KIAA1008, Anti-RP11-342J4.3, Anti-RRP44, Anti-dis3p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

DIS3-N, monoclonal

form

buffered aqueous solution

mol wt

antigen ~109 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5.0 μg/mL using Human HeLa cells.
western blot: 2.5-5.0 μg/mL using Whole extracts of human HEK-293T cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DIS3(22894)
mouse ... Dis3(72662)
rat ... Dis3(306103)

General description

Monoclonal Anti-DIS3 (mouse IgG1 isotype) is derived from the hybridoma DIS3-N produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. DIS3 homolog, exosome endoribonuclease and 3′-5′ exoribonuclease (DIS3), mitotic control homolog, also known as RRP44 and EXOSC11, is a catalytic component of the eukaryotic RNA exosome. The RNA exosome is a ribonucleolytic complex of 400 kDa. It is composed of a nine catalytically inert subunit core. The exosome is a nuclear complex, also localized to the cytoplasm. DIS3 is predominantly found in the nucleus, whereas DIS3L is mainly cytoplasmic.

Immunogen

Synthetic peptide corresponding to an internal sequence of human DIS3, conjugated to KLH. The corresponding sequence is identical in monkey, pig and dog, and differs by a single amino acid in mouse and rat DIS3.

Application

Monoclonal Anti-DIS3 antibody produced in mouse has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

The RNA exosome is essential for RNA processing and turnover. Its core subunits possess structural function and is associated with substrate recognition. The exosome was characterized as a multiunit complex important for the 3′ → 5′ processing and degradation of many types of RNAs including mRNA, rRNA, snRNA, snoRNA, and tRNA. DIS3 homolog, exosome endoribonuclease and 3′-5′ exoribonuclease (DIS3) and DIS3L are potential exonucleases. DIS3 also displays endonuclease activity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tristetraprolin-mediated hexokinase 2 expression regulation contributes to glycolysis in cancer cells
Kim DJ, et al.
Molecular Biology of the Cell, 30(5), 542-553 (2019)
Mai-Tram Vo et al.
Oncotarget, 8(26), 41903-41920 (2017-04-15)
Mitochondrial dynamics play critical roles in maintaining mitochondrial functions. Here, we report a novel mechanism for regulation of mitochondrial dynamics mediated by tristetraprolin (TTP), an AU-rich element (ARE)-binding protein. Overexpression of TTP resulted in elongated mitochondria, down-regulation of mitochondrial oxidative
Rafal Tomecki et al.
The EMBO journal, 29(14), 2342-2357 (2010-06-10)
The eukaryotic RNA exosome is a ribonucleolytic complex involved in RNA processing and turnover. It consists of a nine-subunit catalytically inert core that serves a structural function and participates in substrate recognition. Best defined in Saccharomyces cerevisiae, enzymatic activity comes
Dong Jun Kim et al.
Molecular biology of the cell, 30(5), 542-553 (2019-01-17)
Hexokinase 2 (HK2) catalyzes the first step of glycolysis and is up-regulated in cancer cells. The mechanism has not been fully elucidated. Tristetraprolin (TTP) is an AU-rich element (ARE)-binding protein that inhibits the expression of ARE-containing genes by enhancing mRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service