Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200498

Sigma-Aldrich

Monoclonal Anti-SET antibody produced in mouse

clone SET7, tissue culture supernatant

Synonym(s):

Monoclonal Anti-2PP2A, Monoclonal Anti-I2PP2A, Monoclonal Anti-IGAAD, Monoclonal Anti-IPP2A2, Monoclonal Anti-PHAPII, Monoclonal Anti-SET nuclear oncogene, Monoclonal Anti-TAF-I, Monoclonal Anti-TAF-IBETA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

SET7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

flow cytometry: 1:25-1:50/test using A549 cells
western blot: 1:2000-1:4000 using A549 total cell extracts

isotype

IgA

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SET(6418)

General description

Monoclonal Anti-SET (mouse IgA isotype) is derived from the hybridoma SET7 produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. The SET nuclear proto-oncogene (SET7) protein localizes both to the nucleus and the cytoplasm.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminus of human SET, conjugated to KLH. The isotype is determined by ELISA using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-SET antibody produced in mouse has been used in immunoblotting and flow cytometry.

Biochem/physiol Actions

SET nuclear proto-oncogene (SET7) is involved in the regulation of signal transduction in normal and cancer cells. It contains a highly acidic C-terminal region that is involved in chromatin remodeling. In fact, SET itself is also a potent and specific inhibitor of protein phosphatase 2A (PP2A) activity and exerts its activity via its N-terminal region. SET also acts as an inhibitor of the tumor suppressor NM23-H1, which is activated by granzyme A DNAse and negatively controls histone acetylation. Interestingly, in acute non-lymphocytic myeloid leukemia, SET was found to be fused to CAN (nucleoporin Nup214), apparently as a result of a chromosomal translocation. As such, the formation of this SET-CAN fusion protein may impair the normal regulation of PP2A and contribute to leukemogenesis. SET was also found to regulate human NK-cell cytotoxicity, by regulating NK-cell IFN-g production and granzyme B gene expression via PP2A inactivation.

Physical form

Solution containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein phosphatase 2A: a highly regulated family of serine/threonine phosphatases implicated in cell growth and signalling.
Janssens V and Goris J
The Biochemical Journal, 353(Pt 3), 417-417 (2001)
Regulation of histone acetylation and transcription by INHAT, a human cellular complex containing the set oncoprotein
Seo SB, et al.
Cell, 104(1), 119-130 (2001)
The PP2A inhibitor SET regulates granzyme B expression in human natural killer cells
Trotta R, et al.
Blood, 117(8), 2378-2384 (2011)
Zusen Fan et al.
Cell, 112(5), 659-672 (2003-03-12)
Granzyme A (GzmA) induces a caspase-independent cell death pathway characterized by single-stranded DNA nicks and other features of apoptosis. A GzmA-activated DNase (GAAD) is in an ER associated complex containing pp32 and the GzmA substrates SET, HMG-2, and Ape1. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service