Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200496

Sigma-Aldrich

Anti-LMOD1 antibody, Mouse monoclonal

clone LMOD1-12, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-LMOD1 antibody produced in mouse, Monoclonal Anti-SM-LMOD, Monoclonal Anti-leiomodin 1 (smooth muscle)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

LMOD1-12, monoclonal

form

buffered aqueous solution

mol wt

~70 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

western blot: 0.1-0.2 μg/mL using whole extracts of HEK-293T cells overexpressing LMOD1.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMOD1(25802)

General description

Monoclonal Anti-LMOD1 (mouse IgG2a isotype) is derived from the hybridoma LMOD1-12 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Leiomodin 1 (LMOD1) shares its homology greatly with tropomodulin, a tropomyosin-binding, actin-capping protein. Leiomodin contains two actin-binding sites, a flexible N-terminal domain with a tropomyosin-binding site and the leucine-rich repeat domain. It also has a C-terminal domain with a short polyproline region and an actin-binding Wiskott-Aldrich syndrome protein (WASP)-homology 2 (WH2) domain. It is expressed predominantly in smooth muscle. The LMOD1 gene is mapped to the human chromosome location 1q32.1.

Specificity

Monoclonal Anti-LMOD1 recognizes human and rat LMOD1.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal region of human LMOD1, conjugated to KLH. The corresponding sequence is identical in mouse, rat, monkey, bovine and canine LMOD1.

Application

Monoclonal Anti-LMOD1 antibody produced in mouse may be used in immunoprecipitation and immunoblotting.

Biochem/physiol Actions

Leiomodin 1 (LMOD1) is an actin filament nucleator in muscle cells. It is found near the pointed ends of the actin filaments and is involved in sarcomere assembly and organization. LMOD1 knockdown results in sarcomere disruption. Overexpression of LMOD1 may be associated with Graves′ disease and thyroid-associated ophthalmopathy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional regulatory mechanism of smooth muscle cell-restricted LMOD1 coronary artery disease locus
Nanda V, et al.
PLoS Genetics, 14(11), e1007755-e1007755 (2018)
Leiomodin is an actin filament nucleator in muscle cells
Chereau D, et al.
Science (New York, N.Y.), 320(5873), 239-243 (2008)
Identification of potential ligand molecule and drug docking studies on autoimmune thyroid disease (Human) using chem informatics techniques
Agalyaa S and Balaji M
International Journal of Nanoscience, 3(4), 72-77 (2013)
Leiomodin/tropomyosin interactions are isoform specific
Kostyukova AS
Archives of Biochemistry and Biophysics, 465(1), 227-230 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service