Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200492

Sigma-Aldrich

Anti-Adrenomedullin (N-terminal) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ADM, Anti-AM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1,000 using formalin-fixed, paraffin-embedded human small intestine.
indirect ELISA: 1:10,000-1:20,000 using human adrenomedullin.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADM(133)

General description

Adrenomedullin is isolated from human pheochromocytoma. It is highly expressed in endothelial cells and vascular smooth muscle cells and is localized in highly vascularized tissues including kidney, intestine, stomach, brain, lung, heart and placenta.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminus of human adrenomedullin (ADM), conjugated to KLH. The corresponding sequence has significant homology (75% sequence identity) in rat and mouse ADM.

Application

Anti-Adrenomedullin (N-terminal) antibody produced in rabbit has been used in immunohistochemistry and enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Adrenomedullin (ADM, AM) is a potent hypotensive peptide. In addition to its vasodilator effect, ADM also exerts a wide range of biological actions including diuretic and cardiotonic effects. ADM is involved in the regulation of hormone release, inflammation, oxidative stress as well as the proliferation, migration and differentiation of various cell types. ADM is thought to play a significant role in angiogenesis and in pathological conditions including cardiovascular disease, ischemia and carcinogenesis. ADM is broadly distributed in many tissues during development and in adulthood. ADM exerts its effects via G-protein coupled calcitonin-like receptors (CRLR) complexed with either receptor activity-modifying protein-2 (RAMP2) or RAMP3.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Receptor Activity-Modifying Proteins: RAMPing up Adrenomedullin Signaling
Gibbons C, et al.
Molecular Endocrinology, 21(4), 783-796 (2007)
Immunohistological localization and possible functions of adrenomedullin
Marutsuka K, et al.
Hypertension Research, 26(Suppl), S33-S40 (2003)
Distribution of adrenomedullin-like immunoreactivity in the rat central nervous system by light and electron microscopy
Serrano J, et al.
Brain Research, 853(2), 245-268 (2000)
The GPCR modulator protein RAMP2 is essential for angiogenesis and vascular integrity
Ichikawa SY, et al.
The Journal of Clinical Investigation, 118(1), 29-39 (2008)
A review of the biological properties and clinical implications of adrenomedullin and proadrenomedullin N-terminal 20 peptide (PAMP), hypotensive and vasodilating peptides
Et T.
Peptides, 22(11), 1693-1711 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service