Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200488

Sigma-Aldrich

Anti-RASD1 (C-terminal) antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-AGS1, Anti-DEXRAS1, Anti-Dexamethasone-induced Ras-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~32 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using cell lysates of HEK-293T overexpressing human RASD1.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Ras-related dexamethasone-induced 1 (RASD1) is a member of the family of Ras monomeric G-proteins and was initially known as a dexamethasone inducible gene. RASD1 consists of four highly conserved guanosine-5′-triphosphate (GTP) binding and hydrolysis pockets (Σ1-Σ4) and also an effector loop. The Σ1 and Σ2 pockets comprise phosphate-magnesium binding domains and the Σ3 and Σ4 consists of guanine nucleotide binding loops. The C-terminal region of RASD1 has a cationic CAAX-domain. It is expressed in the heart, white adipose tissues, liver and kidneys.

Specificity

Anti-RASD1(C-terminal) specifically recognizes human RASD1.

Immunogen

synthetic peptide corresponding to a sequence near the C-terminus of rat RASD1, conjugated to KLH. The corresponding sequence is highly conserved in human RASD1 (83% sequence identity) and in rat RASD1 (94% sequence identity).

Application

Anti-RASD1 (C-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Ras-related dexamethasone-induced 1 (RASD1) /Dexras1 regulates several signaling pathways such as N-methyl-d-aspartic acid receptors (NMDAR)-nitric oxide, G-protein coupled receptors (GPCRs), mitogen-activated protein kinase/extracellular signal-regulated protein kinase (MAPK/ERK) and protein kinase C (PKC). Dexras1 is S-nitrosylated and activated by nitric oxide (NO) donors and by NMDA receptor-stimulated NO-synthesis in cortical neurons. It acts as a novel physiological target of neuronal NO synthase (nNOS). The formation of a ternary complex between nNOS, the nNOS adaptor protein CAPON and DexRAS1 has been shown to enhance the ability of nNOS to activate Dexras1. Knock-out of nNOS in mice results in a selective decrease of Dexras1 activation. NMDA receptor-stimulated NO-signaling has been shown to mediate neuronal iron homeostasis via Dexras1. An iron-uptake signaling cascade including NMDA-NO-Dexras1-peripheral benzodiazepine receptor associated protein (PAP7)-divalent metal transporter 1 (DMT1) is thought to mediate NMDA neurotoxicity. Dexras1 interacts with FE65 and can direct modulate FE65-amyloid precursor protein (APP)-mediated transcription.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °Cfor up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation beforeuse. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RASD1
Bouchard-Cannon P and Cheng HYM
Encyclopedia of Signaling Molecules, 1-9 (2017)
Rasd1, a small G protein with a big role in the hypothalamic response to neuronal activation
Greenwood MP, et al.
Molecular Brain, 9(1), 1-17 (2016)
NMDA receptor-nitric oxide transmission mediates neuronal iron homeostasis via the GTPase Dexras1
Cheah JH, et al.
Neuron, 51(4), 431-440 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service