Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB4200486

Sigma-Aldrich

Anti-Dysbindin (C-terminal) antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-DBND, Anti-DTNBP1, Anti-HPS7, Anti-My031, Anti-SDY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

human, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 10-20 μg using extracts of HEK-293T cells overexpressing human dysbindin (DTNBP1).
indirect immunofluorescence: 5-10 μg/mL using PC12 cells.
western blot: 0.5-1 μg/mL using extracts of HEK-293T cells overexpressing human dysbindin (DTNBP1).

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Dysbindin-1 (DTNBP1) is a protein, encoded by the dystrobrevin-binding protein 1 gene. Initially, it was identified to be a constituent of the dystrophin-associated protein complex (DPC) in skeletal muscle cells. The DTNBP1 protein consists of two coiled-coil domains and it exists in three isoforms, namely dysbindin-1A, -1B and -1C. DTNBP1 mRNA is widely expressed in the brain, located in postsynaptic densities and synaptic vesicles of neurons. The gene is present at the human chromosome location 6p22.3.

Specificity

Anti-Dysbindin(C-terminal) specifically recognizes human and rat dysbindin.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminus of human dysbindin, conjugated to KLH. The antibody is affinity-purified using the immunizing peptide immobilized on agarose.

Application

Anti-Dysbindin (C-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Dysbindin (also known as DTNBP1) is a neuronal protein that controls synaptic homeostasis by regulating the exocytosis and formation of synaptic vesicles. Polymorphism of DTNBP1 confers susceptibility to schizophrenia through a decreased expression of dysbindin, especially in the hippocampal formation. Deletion mutation of DTNBP1 in sandy (sdy) mouse is linked to impaired long term memory retention and working memory. Mutations in the human DTNBP1 gene are associated with Hermansky-Pudlak syndrome (HPS). Dysbindin is a key component of the biogenesis of lysosome-related organelles complex 1 (BLOC-1), which regulates the trafficking of proteins in the lysosomal pathway. At postsynaptic densities, it is involved in trafficking and tethering of receptors and signal transduction proteins.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H ermansky-P udlak syndrome: pigmentary and non-pigmentary defects and their pathogenesis
Wei AH and Li W
Pigment Cell & Melanoma Research, 26(2), 176-192 (2013)
Dysbindin-1 involvement in the etiology of schizophrenia
Wang H, et al.
International Journal of Molecular Sciences, 18(10), 2044-2044 (2017)
Is the dysbindin gene (DTNBP1) a susceptibility gene for schizophrenia?
Williams NM, et al.
Schizophrenia Bulletin, 31(4), 800-805 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service