Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200482

Sigma-Aldrich

Anti-ADAR1 antibody, Mouse monoclonal

clone ADAR61, purified from hybridoma cell culture

Synonym(s):

Anti-ADAR1, Anti-DRADA, Anti-DSH, Anti-DSRAD, Anti-G1P1, Anti-IFI4, Anti-K88DSRBP, Anti-P136, Monoclonal Anti-ADAR1 antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ADAR61, monoclonal

form

buffered aqueous solution

mol wt

antigen 80-130 kDa

species reactivity

monkey, human, rat

concentration

~1.0 mg/mL

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: 2.0-4.0 μg/mL using U2OS total cell extracts

isotype

IgG3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAR(103)

General description

Monoclonal Anti-ADAR1 (mouse IgG3 isotype) is derived from the hybridoma ADAR61 produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. Adenosine deaminase that acts in RNA 1 (ADAR1) and ADAR2 are expressed in most tissues but in general, the pre-mRNA being edited represents receptors of the central nervous systems. ADAR1 has several isoforms which include ADAR1 (150kDa) and two short isoforms (80 and 110 kDa) that are functional.

Immunogen

a peptide corresponding to a sequence at the C-terminus of human ADAR, conjugated to KLH.

Application

Monoclonal Anti-ADAR1 antibody produced in mouse has been used in:
  • immunoblotting
  • immunoprecipitation
  • flow cytometry

Biochem/physiol Actions

RNA editing by hydrolytic deamination of adenosine (A) to inosine (I) in double-stranded (ds)RNA is the most common type of editing in higher eukaryotes. This RNA editing event is catalyzed by the adenosine deaminase that acts in RNA (ADAR) enzyme. Two ADAR enzymes have been shown to possess enzymatic activity in mammals, ADAR1 and ADAR2. Both can convert specific adenosine to inosine in pre-mRNA and can also convert up to 40-50% of the adenosines in long synthetic duplex RNAs. ADAR1 plays critical roles in differentiating cells of embryo and adult tissues, supporting cell survival and permitting their further differentiation and maturation. Its role in embryonic development is especially demonstrated within the hematopoietic lineage as well as in adult hematopoietic progenitor cells (HPCs). ADAR1 is also an essential regulator of hematopoietic stem cell maintenance and suppressor of interferon signaling that may protect organisms from the deleterious effects of interferon activation associated with many pathological processes, including chronic inflammation, autoimmune disorders and cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RNA editing as a therapeutic target for CNS disorders
Morabito MV and Emeson RB
Neuropsychopharmacology, 34(1), 246-246 (2009)
The many roles of an RNA editor
Keegan LP, et al.
Nature Reviews. Genetics, 2(11), 869-869 (2001)
RNA editing catalyzed by ADAR1 and its function in mammalian cells
Wang Q
Biochemistry (Moscow), 76(8), 900-900 (2011)
ADAR1 is essential for the maintenance of hematopoiesis and suppression of interferon signaling
Hartner JC, et al.
Nature Immunology, 10(1), 109-109 (2009)
Editor meets silencer: crosstalk between RNA editing and RNA interference
Nishikura K
Nature Reviews in Molecular and Cell Biology, 7(12), 919-919 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service