SAB4200474
Anti-DIS3 antibody, Mouse monoclonal
clone DIS3-3, purified from hybridoma cell culture
Synonym(s):
Anti-EXOSC11, Anti-KIAA1008, Anti-RP11-342J4.3, Anti-RRP44, Anti-dis3p, Monoclonal Anti-DIS3 antibody produced in mouse
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
DIS3-3, monoclonal
form
buffered aqueous solution
mol wt
antigen ~109 kDa
species reactivity
human
concentration
~1.0 mg/mL
technique(s)
immunoprecipitation (IP): suitable
indirect immunofluorescence: 2.5-5.0 μg/mL using human HeLa cells
western blot: 1-2 μg/mL using whole extracts of human HEK-293T cells
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... DIS3(22894)
General description
Immunogen
Application
- immunoblotting
- immunoprecipitation
- immunofluorescence
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service