Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200472

Sigma-Aldrich

Anti-SOX11 antibody, Mouse monoclonal

clone SOX11-2, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-SOX11 antibody produced in mouse, Monoclonal Anti-SRY (sex determining region Y)-box 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SOX11-2, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.25-0.5 μg/mL using whole extracts of HEK-293T cells overexpressing human SOX11.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX11(6664)

General description

Monoclonal Anti- SOX11 (mouse IgG1 isotype) is derived from the hybridoma SOX11-2 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Sex determining region Y (SRY)-related high-mobility-group (HMG) box 11 (SOX11) protein is a member of the SOX family of transcription factors. It is expressed in human and mouse fetuses in the lungs, peripheral and central nervous system and gastrointestinal tract. The SOX11 gene is mapped to human chromosome location 2p25.2. SOXC proteins comprise a transactivation domain in the C-terminal region and a highly conserved HMG box DNA-binding domain in the N-terminal.

Specificity

Monoclonal Anti- SOX11 recognizes human SOX11.

Immunogen

synthetic peptide corresponding to an internal region of human SOX11, conjugated to KLH. The corresponding sequence is identical in monkey and pig and differs by 3 amino acids in mouse and rat.

Application

Monoclonal Anti-SOX11 antibody produced in mouse may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Sex determining region Y (SRY)-related high-mobility-group (HMG) box 11 (SOX11) protein plays a role in tumorigenesis. It regulates embryonic development of the central nervous system. It also plays a major role in the determination of cell fate. SOX11 modulates progenitor and stem cell behavior. Dysregulation of SOX11 is associated with neurodevelopmental disorders, osteoarthritis and a wide variety of cancers.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulatory roles for SOX11 in development, stem cells and cancer
Tsang SM, et al.
Seminars in Cancer Biology (2020)
SOX11: friend or foe in tumor prevention and carcinogenesis?
Yang Z, et al.
Therapeutic Advances in Medical Oncology, 11, 1758835919853449-1758835919853449 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service