Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200467

Sigma-Aldrich

Anti-Migfilin antibody, Mouse monoclonal

clone 43, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-CAL, Monoclonal Anti-FBLIM1, Monoclonal Anti-FBLP-1, Monoclonal Anti-FBLP1, Monoclonal Anti-Migfilin antibody produced in mouse, Monoclonal Anti-filamin binding LIM protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

43, monoclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 20 μg/mL using A431 cells.
western blot: 0.1-0.2 μg/mL using A431 total cell extracts.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBLIM1(54751)

General description

FBLIM1 (filamin binding LIM protein 1) is present at the cell-cell adhesion areas. The protein has the amino-terminal, proline rich-region and carboxy-terminal LIM (LIN-11, ISL-1 and MEC-3) domains.[1] It is commonly referred to as migfilin.
Monoclonal Anti-Migfilin (mouse IgG1 isotype) is derived from the hybridoma 43 produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. The Migfilin gene is mapped to the human chromosome location 1p36.21.

Specificity

Monoclonal Anti-Migfilin recognizes human migfilin.

Immunogen

recombinant human migfilin, fusion protein.1 The isotype is determined by ELISA using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-Migfilin antibody produced in mouse may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry
  • immunohistochemistry

Biochem/physiol Actions

FBLIM1 (filamin binding LIM protein 1) plays an important role in association between the cell membrane and the actin cytoskeleton. It mainly interacts with MIG-2 (mitogen-inducible gene 2 protein), filamin and VASP (vasodilator-stimulated phosphoprotein), thus controlling cell shape and movements. In cardiomyocytes, it interacts with transcription factor CSX/NKX2-5 (cardiac-specific homeobox) and enhances the differentiation. The FBLIM1 gene is upregulated in osteoarthritis chondrocytes and might be involved with osteoarthritis pathogenesis. In various carcinoma cells, the FBLIM1-associated signaling in disrupted which leads to abnormal Src activation and anoikis resistance in cells. This gene is downregulated in breast cancer cells. In esophageal cancer cells, it suppresses invasion partly by enhancing degradation of β-catenin. However, in glioma cells FBLIM1 promotes migration as well as invasion.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

circFBLIM1 act as a ceRNA to promote hepatocellular cancer progression by sponging miR-346
Bai N, et al.
Journal of Experimental & Clinical Cancer Research, 37(1), 172-172 (2018)
Mitali Das et al.
PloS one, 6(10), e26355-e26355 (2011-11-02)
Cell adhesion and migration depend on engagement of extracellular matrix ligands by integrins. Integrin activation is dynamically regulated by interactions of various cytoplasmic proteins, such as filamin and integrin activators, talin and kindlin, with the cytoplasmic tail of the integrin
Migfilin's elimination from osteoarthritic chondrocytes further promotes the osteoarthritic phenotype via ?-catenin upregulation.
Gkretsi V, et al.
Biochemical and Biophysical Research Communications, 430, 494-499 (2013)
Migfilin interacts with Src and contributes to cell-matrix adhesion-mediated survival signaling.
Zhao J, et al.
The Journal of Biological Chemistry, 284, 34308-34320 (2009)
Migfilin regulates esophageal cancer cell motility through promoting GSK-3?-mediated degradation of ?-catenin.
He H, et al.
Molecular Cancer Research, 10, 273-281 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service