SAB4200457
Anti-EXOSC7 (N-terminal) antibody produced in rabbit
~1.0 mg/mL, affinity isolated antibody
Synonym(s):
Anti-EAP1, Anti-Exosome complex component RRP42, Anti-Exosome complex exonuclease RRP42, Anti-Exosome component 7, Anti-RRP42, Anti-Ribosomal RNA-processing protein 42, Anti-Rrp42p, Anti-hRrp42p, Anti-p8
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~34 kDa
species reactivity
human
concentration
~1.0 mg/mL
technique(s)
immunoblotting: 0.5-1.0 μg/mL using whole extracts of human HeLa cells.
immunoblotting: 1-2 μg/mL using whole extracts of mouse NIH-3T3.
immunoblotting: 2-4 μg/mL using whole extracts of rat A10 cells.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... EXOSC7(23016)
General description
Specificity
Immunogen
Application
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service