Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200447

Sigma-Aldrich

Monoclonal Anti-BTBD9 antibody produced in mouse

clone BTBD9-205, tissue culture supernatant

Synonym(s):

Anti-BTB (POZ) domain containing 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

BTBD9-205, monoclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

mouse, human, monkey, canine

technique(s)

indirect immunofluorescence: 1:500-1:1000 using HeLa cells
western blot: 1:8000-1:16,000 using extracts of K562 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BTBD9(114781)

General description

Monoclonal Anti-BTBD9 (mouse IgM isotype) is derived from the hybridoma BTBD9-205 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide corresponding to a sequence at the C-terminus of human BTBD9 conjugated to KLH protein, that contains one BTB/POZ domain and one BACK (BTB/Kelch associated) domain. The BTB/POZ domain mediates homomeric and heteromeric protein-protein interactions.
The BTBD9 gene is mapped to human chromosome 6p21.2.

Specificity

Monoclonal Anti-BTBD9 recognizes human, monkey, dog and mouse BTBD9.

Immunogen

a peptide corresponding to a sequence at the C-terminus of human BTBD9, conjugated to KLH.

Application

Monoclonal Anti-BTBD9 antibody produced in mouse has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

BTBD9 protein plays a vital role in protein-protein interactions.
Mutations in BTBD9 (BTB/POZ domain-containing protein 9) is associated with restless legs syndrome type 6 (RLS6) and Tourette syndrome.

Physical form

Culture supernatant solution containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 0C in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of intronic variants of the BTBD9 gene with Tourette syndrome
Riviere JB, et al.
Archives of Neurology, 66(10), 1267-1272 (2009)
Update of the pathophysiology of the restless-legs-syndrome
Paulus W, et al.
Movement Disorders, 22(S18), S431-S439 (2007)
Soraya Arzhan et al.
Blood purification, 49(1-2), 222-227 (2019-12-19)
Uremic pruritus (UP) and restless legs syndrome (RLS) are highly prevalent complaints among patients with end-stage renal disease (ESRD) undergoing chronic dialysis. These chronic troublesome symptoms lead to a significant decrease in quality of life (QOL) and increase in mortality
The BTBD9 gene polymorphisms in Polish patients with Gilles de la Tourette syndrome
Janik P, et al.
Acta Neurobiologiae Experimentalis, 74, 218-226 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service