Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200444

Sigma-Aldrich

Anti-Aurka/Aurora-A Kinase antibody, Mouse monoclonal

~1.0 mg/mL, clone AR-7, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-AIK, Monoclonal Anti-ARK1, Monoclonal Anti-AURA, Monoclonal Anti-AURORA2, Monoclonal Anti-Aurka/Aurora-A Kinase antibody produced in mouse, Monoclonal Anti-BTAK, Monoclonal Anti-PPP1R47, Monoclonal Anti-STK15, Monoclonal Anti-STK6, Monoclonal Anti-STK7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

AR-7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~46 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5.0 μg/mL using HT-29 cells
western blot: 2-4 μg/mL using extracts of THP-1 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AURKA(6790)

General description

Monoclonal Anti-Aurka/Aurora-A Kinase (mouse IgG1 isotype) is derived from the hybridoma AR-7 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Aurka/Aurora-A Kinase is a member of serine/threonine kinase family. The AURKA gene is mapped on the human chromosome at 20q13.2.

Specificity

Monoclonal Anti-Aurka/Aurora-A Kinase recognizes human and monkey Aurka.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminus of human Aurka, conjugated to KLH. The isotype is determined by ELISA using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-Aurka/Aurora-A Kinase antibody produced in mouse may be used in immunoblotting and immunocytochemistry.

Biochem/physiol Actions

Aurka/Aurora-A Kinase is involved in correct centrosomal and microtubule functions. It is also involved in the chromosomal segregation during normal cellular mitosis. This protein plays a role in the recruitment of cyclin-dependent kinase (CDK)-cyclin B1 to the centrosome, which correlates with its activation and the fidelity of the cells to mitosis. Ectopic expression and altered activity of the AURKA gene are observed to induce centrosome amplification leading to genomic instability, aneuploidy and abnormal mitosis. Overexpression of the AURKA gene is observed in a spectrum of cancers such as colorectal, breast and gastric cancers. Overexpression of this gene is also associated with high-grade tumors, invasion and metastasis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing or storage in “frost-free” freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anke H Sillars-Hardebol et al.
Gut, 61(11), 1568-1575 (2011-12-31)
Progression of a colorectal adenoma to invasive cancer occurs in a minority of adenomas and is the most crucial step in colorectal cancer pathogenesis. In the majority of cases, this is associated with gain of a substantial part of chromosome
Jean Yves Cremet et al.
Molecular and cellular biochemistry, 243(1-2), 123-131 (2003-03-07)
We have developed monoclonal antibodies against the human aurora-A serine/threonine kinase. After immunization of a mouse, a fusion was performed to obtain hybridomas that were selected because they produced immunoglobulin positively reacting against the protein used for immunization. We isolated
A Krystyniak et al.
Oncogene, 25(3), 338-348 (2005-09-15)
Mitotic kinases are the ultimate target of pathways sensing genotoxic damage and impinging on the cell cycle machinery. Here, we provide evidence that Aurora A (AurA) was inhibited upon generation of double-strand breaks in DNA. We demonstrate that AurA was
Stéphanie Dutertre et al.
Journal of cell science, 117(Pt 12), 2523-2531 (2004-05-07)
Aurora-A protein kinase, which is the product of an oncogene, is required for the assembly of a functional mitotic apparatus and the regulation of cell ploidy. Overexpression of Aurora-A in tumour cells has been correlated with cancer susceptibility and poor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service