Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200442

Sigma-Aldrich

Anti-COG6 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-COD2, Anti-Component of oligomeric golgi complex 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of human HeLa cells
western blot: 2-4 μg/mL using whole extracts of human HEK-293T cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG6(57511)

General description

Conserved Oligomeric Golgi complex 6 (COG6), also known as COD2, is a member of the conserved oligomeric Golgi (COG) complex. COG complex is composed of eight distinct subunits organized in two heterotrimeric groups, Cog2-Cog3- Cog4 and Cog5-Cog6-Cog7, which are linked by the dimeric group formed by Cog1 and Cog8.

Immunogen

a peptide corresponding to the C-terminal region of human COG6, conjugated to KLH. The corresponding sequence differs by three amino acids in mouse and rat COG6.

Application

Anti-COG6 (C-terminal) antibody produced in rabbit has been used in various immunochemical techniques including immunoblotting and immunoprecipitation.

Biochem/physiol Actions

COG complex is an evolutionarily conserved multi-subunit protein complex that regulates membrane trafficking and maintenance of Golgi glycosylation machinery in eukaryotic cells. COG complex interacts directly with the target membrane (t-SNARE) Syntaxin 6 through COG6 subunit. COG6-depleted cells display reduced steady-state level of Syntaxin 6 and attenuated endosome-to- trans‐Golgi network (TGN) retrograde trafficking. Deficiency of COG6 causes a combined N- and Oglycosylation deficiency of the congenital disorders of glycosylation, designated as CDG-IIL (COG6- congenital disorders of glycosylation (CDG)).

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maintaining order: COG complex controls Golgi trafficking, processing, and sorting
Blackburn JB, et al.
Febs Letters, 593(17), 2466-2487 (2019)
The COG complex interacts directly with Syntaxin 6 and positively regulates endosome-to-TGN retrograde transport
Laufman O, et al.
The Journal of cell biology, 194, 459-472 (2011)
Fatal outcome due to deficiency of subunit 6 of the conserved oligomeric Golgi complex leading to a new type of congenital disorders of glycosylation
Lubbehusen J, et al.
Human Molecular Genetics, 19, 3623-3633 (2010)
Orly Laufman et al.
The Journal of cell biology, 194(3), 459-472 (2011-08-03)
The conserved oligomeric Golgi (COG) complex has been implicated in the regulation of endosome to trans-Golgi network (TGN) retrograde trafficking in both yeast and mammals. However, the exact mechanisms by which it regulates this transport route remain largely unknown. In
Jürgen Lübbehusen et al.
Human molecular genetics, 19(18), 3623-3633 (2010-07-08)
Deficiency of subunit 6 of the conserved oligomeric Golgi (COG6) complex causes a new combined N- and O-glycosylation deficiency of the congenital disorders of glycosylation, designated as CDG-IIL (COG6-CDG). The index patient presented with a severe neurologic disease characterized by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service