Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200440

Sigma-Aldrich

Anti-COG5 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-13S Golgi transport complex 90 kDa subunit, Anti-COG complex subunit 5, Anti-Component of oligomeric Golgi complex 5, Anti-Conserved oligomeric Golgi complex subunit 5, Anti-GTC-90, Anti-Golgi transport complex 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~100 kDa

species reactivity

human, rat, mouse

concentration

~1.0 mg/mL

technique(s)

western blot: 2-4 μg/mL using whole extracts of HEK-293T cells overexpressing human COG5.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG5(10466)
mouse ... Cog5(238123)
rat ... Cog5(298945)

Looking for similar products? Visit Product Comparison Guide

General description

Conserved oligomeric Golgi complex 5 (COG5), also known as CDG2I, GTC90 and GOLTC1, is a member of the conserved oligomeric Golgi (COG) complex. The complex is an evolutionarily conserved multisubunit protein COG complex. COG is composed of eight distinct subunits organized in two heterotrimeric groups, Cog2-Cog3-Cog4 and Cog5-Cog6-Cog7, which are linked by the dimeric group formed by Cog1 and Cog8.

Immunogen

peptide corresponding to the C-terminal region of human COG5, conjugated to KLH. The corresponding sequence is identical in mouse, rat, canine and monkey COG5.

Application

Anti-COG5 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Conserved Oligomeric Golgi complex 5 (COG5) complex that regulates membrane trafficking and maintenance of Golgi glycosylation machinery in eukaryotic cells. Mutations in COG5 gene result in congenital disorder of glycosylation type 2I.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conserved oligomeric Golgi complex specifically regulates the maintenance of Golgi glycosylation machinery
Pokrovskaya ID, et al.
Glycobiology, 21(12), 1554-1569 (2011)
Sami Tabbarah et al.
Scientific reports, 10(1), 21269-21269 (2020-12-06)
Leber congenital amaurosis (LCA), a form of autosomal recessive severe early-onset retinal degeneration, is an important cause of childhood blindness. This may be associated with systemic features or not. Here we identified COG5 compound-heterozygous variants in patients affected with a
Comparative analyses of the Conserved Oligomeric Golgi (COG) complex in vertebrates
Quental R, et al.
BMC Evolutionary Biology, 10(1), 212-212 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service