Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200432

Sigma-Aldrich

Anti-RASD2 antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-RAS homolog enriched in striatum, Anti-RASD family, member 2, Anti-RHES, Anti-TEM-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~34 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2-4 μg using lysates of HEK-293T cells overexpressing human RASD2
indirect immunofluorescence: 5-10 μg/mL using HEK-293T cells overexpressing RASD2
western blot: 0.5-1 μg/mL using lysates of HEK-293T cells overexpressing human RASD2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RASD2(23551)

General description

RASD2 (Ras homolog enriched in striatum) is a member of the family of Ras small G proteins. RASD2 is a 266 amino acid GTP binding protein, which is expressed in the striatum. The gene is located on human chromosome 22 (q12.3).

Immunogen

synthetic peptide corresponding to a sequence near the C-terminus of human RASD2, conjugated to KLH. The corresponding sequence is highly conserved (83% sequence identity) in rat and mouse RASD2.

Application

Anti-RASD2 antibody produced in rabbit has been used in:
  • immunoblotting
  • immunofluorescence
  • immunoprecipitation

Biochem/physiol Actions

RASD2 (Ras homolog enriched in striatum) prevents the activation of Gαs and promotes dopamine D2 receptor-mediated activation of Gαi/o. It mediates mammalian target of rapamycin (mTOR) signaling and L-DOPA(L-3,4-dihydroxyphenylalanine)-induced dyskinesia. It is suggested to be the principal determinant of striatal mTOR activation. RASD2 is associated with Huntington′s disease (HD). Rhes-mHtt interaction is implicated in neuropathology of HD in the striatum.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhes, a striatal-selective protein implicated in Huntington disease, binds beclin-1 and activates autophagy
Mealer RG, et al.
Test (2014)
Rasd2 modulates prefronto-striatal phenotypes in humans and `schizophrenia-like behaviors? in mice
Vitucci D, et al.
Neuropsychopharmacology, 41(3), 916-916 (2016)
Rhes, a striatal-selective protein implicated in Huntington disease, binds beclin-1 and activates autophagy
Mealer RG, et al.
The Journal of Biological Chemistry, 289(6), 3547-3554 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service