Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200428

Sigma-Aldrich

Anti-COG8 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CDG2H, Anti-COG complex subunit 8, Anti-Component of oligomeric golgi complex 8, Anti-Conserved oligomeric Golgi complex subunit 8, dependent on RIC1, Anti-Conserved oligomeric golgi complex component 8, Anti-DOR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of human HEK-293T cells
western blot: 2.5-5 μg/mL using whole extracts of HEK-293 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG8(84342)

General description

Conserved oligomeric Golgi complex 8 (COG8) belongs to the COG complex. COG complex is a conserved multi-subunit protein complex. COG complex is composed of eight distinct subunits organized in two heterotrimeric groups, COG-2,-3,-4 and COG-5,-6,-7, which are linked via a dimeric group formed by COG1 and COG8. COG8 is located on the human chromosome at 16q22.1.

Specificity

Anti-COG8 (N-terminal) recognizes human COG8.

Immunogen

a peptide corresponding to the N-terminal region of human COG8, conjugated to KLH. The corresponding sequence differs by a single amino acid in rat and mouse COG8.

Application

Anti-COG8 (N-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Conserved oligomeric Golgi (COG) complex regulates membrane trafficking and maintenance of Golgi glycosylation machinery in eukaryotic cells. Mutations in the COG8 gene causes congenital disorder of glycosylation, type IIh, characterized by under-glycosylated serum proteins. The symptoms include severe psychomotor retardation, failure to thrive, seizures, and dairy and wheat product intolerance.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Kranz et al.
Human molecular genetics, 16(7), 731-741 (2007-03-03)
We describe a new Type II congenital disorder of glycosylation (CDG-II) caused by mutations in the conserved oligomeric Golgi (COG) complex gene, COG8. The patient has severe psychomotor retardation, seizures, failure to thrive and intolerance to wheat and dairy products.
Isabel Pereira-Castro et al.
Human genetics, 131(2), 265-274 (2011-08-02)
Although gene-free areas compose the great majority of eukaryotic genomes, a significant fraction of genes overlaps, i.e., unique nucleotide sequences are part of more than one transcription unit. In this work, the evolutionary history and origin of a same-strand gene
Rita Quental et al.
BMC evolutionary biology, 10, 212-212 (2010-07-17)
The Conserved Oligomeric Golgi (COG) complex is an eight-subunit assembly that localizes peripherally to Golgi membranes and is involved in retrograde vesicular trafficking. COG subunits are organized in two heterotrimeric groups, Cog2, -3, -4 and Cog5, -6, -7, linked by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service