Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200426

Sigma-Aldrich

Anti-COG3 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-COG complex subunit 3, tethering factor SEC34, Anti-SEC34, Anti-component of oligomeric golgi complex 3, Anti-conserved oligomeric Golgi complex subunit 3, Anti-p94, Anti-vesicle-docking protein SEC34 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~94 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extracts of human HEK-293T, mouse NIH-3T3 and rat PC12 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG3(83548)
mouse ... Cog3(338337)
rat ... Cog3(361073)

General description

The conserved oligomeric Golgi complex 3 (COG3) protein is a member of the conserved oligomeric Golgi (COG) complex. COG complex is an evolutionarily conserved multi-subunit protein complex. COG complex contains eight distinct subunits organized in two heterotrimeric groups, COG2-COG3-COG4 and COG5-COG6-COG7, which are linked by the dimeric group formed by COG1 and COG8. The COG3 gene is mapped on the human chromosome at 13q14.13.

Specificity

Anti-COG3 recognizes human, mouse and rat COG3.

Immunogen

a peptide corresponding to an internal region of human COG3, conjugated to KLH. The corresponding sequence is identical in mouse, rat, monkey and bovine COG3.

Application

Anti-COG3 antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Conserved oligomeric Golgi complex 3 (COG3) protein is involved in the endoplasmic reticulum (ER)-Golgi membrane trafficking. COG3 protein depletion hinders vesicle-mediated Golgi retrograde trafficking in HeLa cells and results in the accumulation of COG complex-dependent vesicles in the cell cytoplasm.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey N Zolov et al.
The Journal of cell biology, 168(5), 747-759 (2005-02-25)
The conserved oligomeric Golgi (COG) complex is an evolutionarily conserved multi-subunit protein complex that regulates membrane trafficking in eukaryotic cells. In this work we used short interfering RNA strategy to achieve an efficient knockdown (KD) of Cog3p in HeLa cells.
Caterina Chiappetta et al.
Oncotarget, 8(46), 80416-80428 (2017-11-09)
Osteosarcoma is the most common pediatric primary non-hematopoietic bone tumor. Survival of these young patients is related to the response to chemotherapy and development of metastases. Despite many advances in cancer research, chemotherapy regimens for osteosarcoma are still based on
Rita Quental et al.
BMC evolutionary biology, 10, 212-212 (2010-07-17)
The Conserved Oligomeric Golgi (COG) complex is an eight-subunit assembly that localizes peripherally to Golgi membranes and is involved in retrograde vesicular trafficking. COG subunits are organized in two heterotrimeric groups, Cog2, -3, -4 and Cog5, -6, -7, linked by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service