SAB4200422
Anti-Lamin A/C R482W antibody, Mouse monoclonal
clone 5H8-B4, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-LMN1, Monoclonal Anti-LMNA, Monoclonal Anti-Lamin A/C, Monoclonal Anti-Lamin A/C R482W antibody produced in mouse, Monoclonal Anti-Prelamin-A/C
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
5H8-B4, monoclonal
form
buffered aqueous solution
species reactivity
human
concentration
~1.0 mg/mL
technique(s)
western blot: 0.1-0.2 μg/mL using a recombinant protein encoding a fragment of human Lamin A/C containing the R482W substitution.
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... LMNA(4000)
General description
Specificity
Immunogen
Application
- immunoblotting[1]
- immunoprecipitation
- immunofluorescence
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service