Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200419

Sigma-Aldrich

Anti-ZFYVE28/LST2 antibody, Mouse monoclonal

clone LST139, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-LST2, Monoclonal Anti-LYST2, Monoclonal Anti-ZFYVE28/LST2 antibody produced in mouse, Monoclonal Anti-zinc finger, FYVE domain containing 28

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

LST139, monoclonal

form

buffered aqueous solution

mol wt

antigen ~95 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: 0.25-0.5 μg/mL using MCF-7 cells.
western blot: 5.0-10.0 μg/mL using extracts of MCF-7 cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFYVE28(57732)

General description

Monoclonal Anti-ZFYVE28/LST2 (mouse IgG1 isotype) is derived from the hybridoma LST139 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide. Zinc finger FYVE domain-containing protein 28 (ZFYVE28) localizes to the early endosome membrane in the absence of monoubiquitination while it localizes to the cytosol when it is monoubiquitinated. The ZFYVE28 gene is located on the human chromosome at 4p16.3.

Specificity

Monoclonal Anti-ZFYVE28/LST2 recognizes human and bovine ZFYVE28/LST2.

Immunogen

peptide corresponding to the N-terminus of human ZFYVE28 /LST2, conjugated to KLH. The corresponding sequence is identical in bovine, and has high homology in dog, rat and mouse ZFYVE28 /LST2.

Application

Monoclonal Anti-ZFYVE28/LST2 antibody produced in mouse may be used in immunoblotting and immunocytochemistry.

Biochem/physiol Actions

Zinc finger FYVE domain-containing protein 28 (ZFYVE28) contains one FYVE-type zinc finger, which mediates the interaction with phosphatidylinositol 3-phosphate (PI3P). ZFYVE28 functions as a negative regulator of epidermal growth factor receptor (EGFR) by promoting its degradation in endosomes when not monoubiquitinated on Lys-87.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcella Zollino et al.
Epilepsia, 55(6), 849-857 (2014-04-18)
Seizure disorder is one of the most relevant clinical manifestations in Wolf-Hirschhorn syndrome (WHS) and it acts as independent prognostic factor for the severity of intellectual disability (ID). LETM1, encoding a mitochondrial protein playing a role in K(+) /H(+) exchange
Yaron Mosesson et al.
Developmental cell, 16(5), 687-698 (2009-05-23)
Genetic screens performed in worms identified major regulators of the epidermal growth factor receptor (EGFR) pathway, including the ubiquitin ligase Cbl/SLI-1. Here we focus on the less-characterized Lst2 protein and confirm suppression of MAPK signals. Unexpectedly, human Lst2, a monoubiquitinylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service