Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200418

Sigma-Aldrich

Monoclonal Anti-KISS1 antibody produced in mouse

~1.0 mg/mL, clone KISS-54, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

KISS-54, monoclonal

form

buffered aqueous solution

mol wt

antigen ~26 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

direct immunofluorescence: 0.25-0.5 μg/mL using MDBK cells
western blot: 1.0-2.0 μg/mL using extracts of MDBK cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KISS1(3814)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-KISS1 (mouse IgG1 isotype) is derived from the hybridoma KISS-54 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide. Kisspeptin is the ligand for G-protein coupled receptor 54 (GPR54). It is expressed in the placenta, hypothalamus and basal ganglia. Kisspeptins are encoded by the gene KISS1 located on the human chromosome at 1q32.1. This protein belongs to the family of arginine-phenylalanine (RF) protein hormones. KISS1 gene is expressed in both central and peripheral tissues.

Specificity

Monoclonal Anti- KISS1 recognizes human, bovine and canine KISS1.

Immunogen

peptide corresponding to a sequence at the C-terminus of human KISS1, conjugated to KLH. The isotype is determined using a double diffusion immunoassay using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-KISS1 antibody produced in mouse may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

KISS1 gene is considered as a human metastasis suppressor gene that can suppress melanoma and breast cancer metastasis, without affecting tumorigenicity. Kisspeptin modulates events downstream of cell-matrix adhesion in mechanisms involving cytoskeletal reorganization. Kisspeptin is involved in initiating the secretion of gonadotropin-releasing-hormone (GnRH) at puberty. This protein functions in the regulation of the hypothalamic-pituitary-gonadal axis, resulting in, influencing fertility and reproduction. KISS1 protein has been suggested as a possible treatment for reproductive disorders as well as clinical treatment of metastases.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °Cinworking aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discardedif not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AXOR12, a novel human G protein-coupled receptor, activated by the peptide KiSS-1
Muir A I, et al.
The Journal of Biological Chemistry, 276(31), 28969-28975 (2001)
Kisspeptin and fertility
Hameed S, et al.
The Journal of Endocrinology, 208(2), 97-97 (2011)
The kisspeptin (KiSS-1)/GPR54 system in cancer biology
Makri A, et al.
Cancer Treatment Reviews, 34(8), 682-692 (2008)
The KISS1 metastasis suppressor: a good night kiss for disseminated cancer cells
Beck B H and Welch D R
European Journal of Cancer, 46(7), 1283-1289 (2010)
Suppression of metastasis in human breast carcinoma MDA-MB-435 cells after transfection with the metastasis suppressor gene, KiSS-1
Lee J H and Welch D R
Cancer Research, 57(12), 2384-2387 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service