Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200417

Sigma-Aldrich

Anti-XPO4 antibody, Mouse monoclonal

clone XP-12, purified from hybridoma cell culture

Synonym(s):

Anti-Exportin 4, Anti-KIAA1721, Monoclonal Anti-Exp4, Monoclonal Anti-XPO4 antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

XP-12, monoclonal

form

buffered aqueous solution

mol wt

antigen ~130 kDa

species reactivity

bovine, chicken, hamster, monkey, canine, human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 1.0-2.0 μg/mL using extracts of HeLa cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPO4(64328)

General description

Monoclonal Anti-XPO4 (mouse IgG1 isotype) is derived from the hybridoma XP-12 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide. Exportin 4 (XPO4) belongs to the karyopherins protein family. It is conserved amongst higher eukaryotes but, lacks obvious orthologues in yeast.

Immunogen

a peptide corresponding to a sequence at the C-terminus of human XPO4.

Application

Monoclonal Anti-XPO4 antibody produced in mouse has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Exportin 4 (XPO4) facilitates the nuclear export of eIF-5A (eukaryotic translation initiation factor 5A). In addition, it also mediates transport Sox proteins, as well as regulates Smad signaling. These functions of XPO4 suggests it to be a candidate tumor suppressor gene. Indeed, its involvement in the progression of human hepatocellular carcinomas (HCC) has been demonstrated, implicating this protein as a potential target for treatment of HCC.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Mechanism of Nuclear Export of Smad3 Involves Exportin 4 and Ran
Kurisaki A, et al.
Molecular and Cellular Biology, 26, 1318-1318 (2006)
Exportin 4 mediates a novel nuclear import pathway for Sox family transcription factors
Gontan C, et al.
The Journal of Cell Biology, 185, 27-27 (2009)
Exportin 4: a mediator of a novel nuclear export pathway in higher eukaryotes
Lipowsky G, et al.
The Embo Journal, 19, 4362-4371 (2000)
Decreased expression of XPO4 is associated with poor prognosis in hepatocellular carcinoma
Liang XT, et al.
Journal of Gastroenterology and Hepatology, 26, 544-549 (2011)
Megumi Tsuchiya et al.
PloS one, 6(10), e25694-e25694 (2011-10-13)
Sox9 is a transcription factor that is required for tissue development in mammals. In general, such transcription factors require co-regulators for precise temporal and spatial control of the activation and inactivation of the numerous genes necessary for precise development during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service