Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200411

Sigma-Aldrich

Anti-non-phospho-TDP-43 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-TAR DNA binding protein 43, Anti-TARDP, Anti-TARDP43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human, mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200 using using mouse kidney
indirect immunofluorescence: 1:200-1:400 using using HeLa cells
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARDBP(23435)
mouse ... Tardbp(230908)

General description

TAR DNA binding protein, TARDP (TDP-43) belongs to the family of heterogeneous nuclear ribonucleoproteins (hnRNPs) that bind single stranded RNA. TDP-43 is predominantly localized to the nucleus. This gene is mapped to human chromosome 1p36.22.

Specificity

Anti-non-phospho-TDP-43 specifically recognizes human and mouse TDP-43 non-phosphorylated on Ser409 and Ser41. Detection of the non-phosphorylated TDP-43 band by immunoblotting is specifically inhibited by the non-phosphorylated TDP-43 immunizing peptide, but not by the corresponding mono-phosphorylated peptides TDP-43 [pSer409] and TDP-43 [pSer410].

Immunogen

synthetic peptide containing non-phosphorylated Ser409 and Ser410 of human TDP-43, conjugated to KLH. The corresponding sequence is identical in mouse TDP-43.

Application

Anti-non-phospho-TDP-43 antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Members of the heterogeneous nuclear ribonucleoprotein (hnRNP) family play multiple roles in the generation and processing of RNA, including transcription, splicing, transport and stability. TAR DNA binding protein, TARDP (TDP-43) has been implicated in the transcription regulation of human immunodeficiency viruses (HIV). TDP-43 has been identified as the major ubiquinated component of cytoplasmic inclusions in frontotemporal lobe degeneration subtype frontotemporal lobar degeneration with ubiquitin-immunoreactive (ub-ir) inclusions (FTLD-U) and amyotrophic lateral sclerosis (ALS).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ALS and FTLD: two faces of TDP-43 proteinopathy
Liscic RM, et al.
European Journal of Neurology, 15(8) (2008)
Heterogeneous Nuclear Ribonucleoprotein E2 (hnRNP E2) Is a Component of TDP-43 Aggregates Specifically in the A and C Pathological Subtypes of Frontotemporal Lobar Degeneration
Kattuah W, et al.
Frontiers in Neuroscience, 13 (2019)
ALS/FTD phenotype in two Sardinian families carrying both C9ORF72 and TARDBP mutations
Chio A, et al.
Journal of Neurology, Neurosurgery, and Psychiatry, 83(7), 730-733 (2012)
TDP-43 regulates retinoblastoma protein phosphorylation through the repression of cyclin-dependent kinase 6 expression
Ayala YM, et al.
Proceedings of the National Academy of Sciences of the USA, 105(10), 3785-3789 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service