Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200409

Sigma-Aldrich

Anti-Dysbindin antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-DBND, Anti-DTNBP1, Anti-HPS7, Anti-My031, Anti-SDY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

rat, mouse, human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using PC12 cells.
western blot: 1-2 μg/mL using HEK-293T cells overexpressing human dysbindin.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Dysbindin (also known as DTNBP1) is a neuronal protein. DTNBP1 mRNA is widely expressed in the brain, located in postsynaptic densities and synaptic vesicles of neurons. Dysbindin-1 is coded by dystrobrevin-binding protein 1 gene (DTNBP1), that is located on the short (p) arm of human chromosome 6.

Immunogen

synthetic peptide corresponding to a sequence at an internal region of human dysbindin, conjugated to KLH. The corresponding sequence is identical in human dysbindin isoforms b, c and in rat and mouse dysbindin.

Application

Anti-Dysbindin antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Dysbindin (DTNBP1) controls synaptic homeostasis by regulating the exocytosis and formation of synaptic vesicles. The DTNBP1 gene has been linked to schizophrenia in humans. Polymorphism of DTNBP1 confers susceptibility to schizophrenia through a decreased expression of dysbindin. Mutations in the human DTNBP1 gene causes a novel form of Hermansky-Pudlak syndrome (HPS) called HPS-7, a genetically heterogeneous disorder caused by abnormal vesicle trafficking to lysosomes and related organelles. Dysbindin, a constituent of the biogenesis of lysosome-related organelles complex 1 (BLOC-1), controls the trafficking of proteins in the lysosomal pathway, suggesting that dysbindin has a role in the biogenesis of lysosome-related organelles.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hermansky-Pudlak syndrome type 7 (HPS-7) results from mutant dysbindin, a member of the biogenesis of lysosome-related organelles complex 1 (BLOC-1)
Li W, et al.
Nature Genetics, 35(1), 84-84 (2003)
The schizophrenia susceptibility gene dysbindin controls synaptic homeostasis
Dickman DK and Davis GW
Science (New York, N.Y.), 326(5956) (2009)
Dysbindin-1 involvement in the etiology of schizophrenia
Wang H, et al.
International Journal of Molecular Sciences, 18(10), 2044-2044 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service