Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200407

Sigma-Aldrich

Anti-phospho-PRAS40 (pSer183) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-AKT1S1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

mouse, human, rat

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 5-10 μg/mL using formalin-fixed, paraffin-embedded human breast carcinoma.
immunoprecipitation (IP): 5-10 μg using extracts of HEK-293T cells.
western blot: 2-4 μg/mL using NIH3T3 and PC12 cell extracts.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer183)

Gene Information

General description

Proline-rich AKT1 substrate 1 (AKT1S1) gene encodes for proline-rich Akt-substrate of 40 kDa (PRAS40) and is located on the human chromosome at 19q13.33.

Specificity

Anti-phospho-PRAS40 (pSer183) specifically recognizes human, mouse and rat phosphorylated PRAS40(pSer183).

Immunogen

synthetic phospho-peptide containing phosphorylated Ser183 of human PRAS40, conjugated to KLH. The corresponding sequence is identical in mouse and rat PRAS40.

Application

Anti-phospho-PRAS40 (pSer183) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunohistochemistry

Biochem/physiol Actions

Proline-rich Akt-substrate of 40 kDa (PRAS40) is a raptor binding protein that inhibits the mammalian target of rapamycin complex 1 (mTORC1) kinase activity. PRAS40 contains a TOR signaling (TOS) motif that mediates its binding to mTORC1. PRAS40 is involved in the protein kinase B (PI3KAkt/PKB) survival pathway. PRAS40 is phosphorylated on multiple residues including Ser-183, Ser-221 and Thr-246 in response to insulin and various growth factors like platelet-derived growth factor (PDGF) and nerve growth factor (NGF), or nutrients like glucose and amino acids. Phosphorylation of PRAS40 by Akt and mTORC1 disrupts the binding between mTORC1 and PRAS40, relieves the inhibitory effect of PRAS40 on mTORC1 activity, and facilitates the binding of PRAS40 to 14-3-3, a cytosolic anchor protein. PRAS40 binds to 14-3-3 with the help of amino acids and insulin which is partially inhibited by rapamycin.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of PRAS40 in Akt and mTOR signaling in health and disease
Wiza C, et al.
American Journal of Physiology. Endocrinology and Metabolism, 302(12), E1453-E1460 (2012)
PRAS40 is a target for mammalian target of rapamycin complex 1 and is required for signaling downstream of this complex
Fonseca BD, et al.
Test, 282(34), 24514-24524 (2007)
Yasemin Sancak et al.
Molecular cell, 25(6), 903-915 (2007-03-28)
The heterotrimeric mTORC1 protein kinase nucleates a signaling network that promotes cell growth in response to insulin and becomes constitutively active in cells missing the TSC1 or TSC2 tumor suppressors. Insulin stimulates the phosphorylation of S6K1, an mTORC1 substrate, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service