Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200404

Sigma-Aldrich

Anti-EXOSC8 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CBP-interacting protein 3, Anti-CIP3, Anti-EAP2, Anti-Exosome component 8,p9, Anti-OIP2, Anti-Opa interacting protein 2, Anti-RP11-421P11.3, Anti-RRP43, Anti-Ribosomal RNA-processing protein 43, Anti-Rrp43p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~34 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of human HEK-293T cells
indirect immunofluorescence: 2.5-5.0 μg/mL using HeLa cells
western blot: 1-2 μg/mL using using whole extracts of HEK-293T cells over-expressing human EXOSC8

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Exosome component 8 (EXOSC8) or ribosomal RNA-processing protein 43 (RRP43), is a 3′ - 5′ exoribonuclease and a component of the eukaryotic exosome, a 400 kDa complex. EXOSC8 is part of the hexameric ring along EXOSC4-7 and EXOSC9. This is further capped by three RNA binding subunits (EXOSC1-EXOSC3). EXOSC8 gene is mapped to human chromosome13q13.3 and is localized in nucleus and cytoplasm.

Specificity

Anti-EXOSC8 (C-terminal) recognizes human EXOSC8.

Immunogen

peptide corresponding to the C-terminal region of human EXOSC8, conjugated to KLH

Application

Anti-EXOSC8 (N-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Exosome component 8 (EXOSC8) interacts with AU-rich element (ARE)-containing mRNAs and its deficiency halts growth in yeast. Mutations in the EXOSC8 gene is implicated in Ponto-cerebellar hypoplasia.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascale Saugier-Veber et al.
European journal of medical genetics, 63(4), 103814-103814 (2019-11-27)
Rhombencephalosynapsis is a rare cerebellar malformation developing during embryogenesis defined by vermian agenesis or hypogenesis with fusion of the cerebellar hemispheres. It occurs either alone or in association with other cerebral and/or extracerebral anomalies. Its association with microlissencephaly is exceedingly
Veronika Boczonadi et al.
Nature communications, 5, 4287-4287 (2014-07-06)
The exosome is a multi-protein complex, required for the degradation of AU-rich element (ARE) containing messenger RNAs (mRNAs). EXOSC8 is an essential protein of the exosome core, as its depletion causes a severe growth defect in yeast. Here we show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service