Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200397

Sigma-Aldrich

Anti-EXOSC4 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Exosome complex component, Anti-Exosome complex exonuclease RRP41, Anti-Exosome component 4, RRP41, Anti-RRP41, Anti-RRP41A, Anti-Rrp41p, Anti-SKI6, Anti-Ski6p, Anti-hRrp41p, Anti-p12A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen ~26 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of human HeLa cells
indirect immunofluorescence: 5-10 μg/mL using human HeLa cells
western blot: 2-4 μg/mL using whole extracts of mouse Hepa1-6 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EXOSC4(54512)
mouse ... Exosc4(109075)

General description

Exosome Component 4 (EXOSC4), also known as RNA-processing protein 41 (RRP41), is a non-catalytic component of the eukaryotic RNA exosome. The eukaryotic exosome is a 400kDa complex composed of a nine subunit catalytically inert core provides structural support and facilitates substrate recognition, and two associated catalytic subunits.
The gene EXOSC4 (exosome component 4) is mapped to human chromosome 8q24.3. The encoded protein is present in the cytoplasm as well as nucleus and has a RNase pleckstrin homology (PH) domain.

Immunogen

peptide corresponding to the N-terminal region of human EXCSC4, conjugated to KLH. The corresponding sequence differs by one amino acid in mouse and rat.

Application

Anti-EXOSC4 antibody produced in rabbit has been used in various immunochemical techniques including immunoblotting, immunoprecipitation and immunofluorescence.

Biochem/physiol Actions

EXOSC4 (exosome component 4) is a core component of the exosome complex and is required for the stability of the complex. The exosome complex exhibits 3′-5′ exoribonuclease function. EXOSC4 lacks ribonuclease activity but is involved in mRNA turnover.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John R Anderson et al.
RNA (New York, N.Y.), 12(10), 1810-1816 (2006-08-17)
We have previously demonstrated that PM-Scl-75, a component of the human exosome complex involved in RNA maturation and mRNA decay, can specifically interact with RNAs containing an AU-rich instability element. Through the analysis of a series of deletion mutants, we
Uttiya Basu et al.
Cell, 144(3), 353-363 (2011-01-25)
Activation-induced cytidine deaminase (AID) initiates immunoglobulin (Ig) heavy-chain (IgH) class switch recombination (CSR) and Ig variable region somatic hypermutation (SHM) in B lymphocytes by deaminating cytidines on template and nontemplate strands of transcribed DNA substrates. However, the mechanism of AID
Erwin L van Dijk et al.
RNA (New York, N.Y.), 13(7), 1027-1035 (2007-06-05)
The human exosome is a 3'-5' exoribonuclease complex that functions both in the nucleus and in the cytoplasm to either degrade or process RNA. Little is known yet about potential differences among core exosome complexes in these different cellular compartments
Chang Ohk Sung et al.
Cancer genetics, 206(5), 145-153 (2013-06-04)
Ovarian clear cell adenocarcinoma (Ov-CCA) is a distinctive subtype of ovarian epithelial carcinoma. In this study, we performed array comparative genomic hybridization (aCGH) and paired gene expression microarray of 19 fresh-frozen samples and conducted integrative analysis. For the copy number
Rafal Tomecki et al.
The EMBO journal, 29(14), 2342-2357 (2010-06-10)
The eukaryotic RNA exosome is a ribonucleolytic complex involved in RNA processing and turnover. It consists of a nine-subunit catalytically inert core that serves a structural function and participates in substrate recognition. Best defined in Saccharomyces cerevisiae, enzymatic activity comes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service