Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200390

Sigma-Aldrich

Anti-HIP1 antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Huntingtin interacting protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~116 kDa

species reactivity

mouse, rat, human

concentration

~1.5 mg/mL

technique(s)

immunoprecipitation (IP): 10 μg using rat brain extracts (S1 fraction).
western blot: 1.5-3.0 μg/mL using mouse brain extracts (S1 fraction).

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIP1(3092)
mouse ... Hip1(215114)
rat ... Hip1(192154)

General description

Huntingtin-interacting protein-1 (HIP1) is a cytosolic protein, which belongs to the huntingtin-interacting protein family. The HIP1 gene is expressed in the endothelium of small blood vessels, gastric epithelium, cardiac muscle, brain tissues and liver. HIP1 and HIP1-related (HIP1R) proteins share the same domain structure, comprising a phospholipid-binding epsin N-terminal homology (ENTH), a leucine zipper motif, a central coiled-coiled and a C-terminal actin-binding domain (THATCH). The gene HIP1 is mapped to human chromosome location 7q11.23.

Specificity

Anti-HIP1 specifically recognizes rat and mouseHIP1.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminus of human HIP1, conjugated to KLH. The corresponding sequence is identical in rat and mouse HIP1.

Application

Anti-HIP1 antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Huntingtin-interacting protein-1 (HIP1) and its homolog HIP1-related (HIP1R) acts as a cofactor in clathrin-mediated membrane traffic and endocytosis. It is implicated in the early recruitment and late-stage maturation of coated pits vesicles. The protein binds to clathrin and the endocytic adaptor complex (AP2) and regulates the binding of HIP1 to actin, leading to alterations in the actin cytoskeleton structure. In addition to its role in endocytosis, HIP1 gene is highly expressed in a variety of human cancers, including colon and prostate cancer. It is also found in the brain, melanoma, breast and ovarian tumors. Overexpression of HIP1 transforms fibroblasts and is associated with increased levels of multiple growth factor receptors. The protein HIP1 is necessary for the survival of germline progenitors and several cell lines. The protein interacts with huntingtin (Htt) and is implicated in the pathogenesis of Huntington′s disease (HD). The fusion between HIP1 and platelet-derived growth factor β receptor is associated with chronic myelomonocytic leukemia.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huntingtin Interacting Protein 1 mutations lead to abnormal hematopoiesis, spinal defects and cataracts
Oravecz-Wilson KI, et al.
Human Molecular Genetics, 13(8), 851 -8867 (2004)
HIP1 exhibits an early recruitment and a late stage function in the maturation of coated pits
Gottfried I, et al.
Cellular and Molecular Life Sciences, 66(17), 2897 -22911 (2009)
Actin binding by Hip1 (huntingtin-interacting protein 1) and Hip1R (Hip1-related protein) is regulated by clathrin light chain
Wilbur JD, et al.
Test, 283(47), 32870 -332879 (2008)
Recurrent distal 7q11. 23 deletion including HIP1 and YWHAG identified in patients with intellectual disabilities, epilepsy, and neurobehavioral problems
Ramocki MB, et al.
American Journal of Human Genetics, 87(6), 857 -8865 (2010)
Huntingtin-interacting protein 1 is overexpressed in prostate and colon cancer and is critical for cellular survival
Rao DS, et al.
The Journal of Clinical Investigation, 110(3), 351 -3360 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service