Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200387

Sigma-Aldrich

Anti-Dysferlin antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Dysferlin antibody produced in rabbit, Anti-Limb girdle muscular dystrophy 2B, Anti-DYSF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~250 kDa

species reactivity

mouse, human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using differentiated C2C12 myoblasts
immunoprecipitation (IP): 5-10 μg using HEK-293T cells over-expressing human dysferlin
western blot: 0.5-1.0 μg/mL using HEK-293T cells over-expressing human dysferlin

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DYSF(8291)
mouse ... Dysf(26903)

General description

Dysferlin is a transmembrane protein that belongs to the ferlin-1 family. It is mostly expressed in the early stages of human development. Dysferlin comprises a C-terminal transmembrane domain and a long N-terminal cytoplasmic region. Dysferlin gene is mapped to 2p13.2.

Specificity

Anti-Dysferlin specifically recognizes human and mouse dysferlin.

Immunogen

synthetic peptide corresponding to an internal sequence of human dysferlin (GeneID: 8291), conjugated to KLH.

Application

Anti-Dysferlin antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Dysferlin localizes in the membrane and aids trafficking along with caveolin protein. The N-terminal region of dysferlin is essential for vesicular traffic and membrane fusion. Mutations in the dysferlin gene is implicated in the limb-girdle muscle dystrophy type 2B (LGMD2B), an autosomal recessive disorder and the related Miyoshi myopathy. It has been suggested to play a role in membrane fusion and repair processes, such as the ability to reseal the sarcolemma upon muscle injury. The integral membrane proteins caveolin-1 and -3 have been shown to regulate the endocytosis of dysferlin.4. Mutations in the caveolin-3 and 1 results in mistargeting and redistribution of dysferlin from the plasma membrane to the Golgi complex.

Physical form

solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolongedstorage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic basis of limb-girdle muscular dystrophies: the 2014 update
Nigro V and Savarese M
Acta Myologica : Myopathies and Cardiomyopathies : Official Journal of the Mediterranean Society of Myology / Edited by the Gaetano Conte Academy for the Study of Striated Muscle Diseases, 33(1), 1-1 (2014)
Caveolin regulates endocytosis of the muscle repair protein, dysferlin
Hernandez-Deviez DJ, et al.
Test, 283(10), 6476-6488 (2008)
Dimple Bansal et al.
Nature, 423(6936), 168-172 (2003-05-09)
Muscular dystrophy includes a diverse group of inherited muscle diseases characterized by wasting and weakness of skeletal muscle. Mutations in dysferlin are linked to two clinically distinct muscle diseases, limb-girdle muscular dystrophy type 2B and Miyoshi myopathy, but the mechanism
S H Laval et al.
Neuropathology and applied neurobiology, 30(2), 91-105 (2004-03-27)
The limb-girdle muscular dystrophies are a diverse group of muscle-wasting disorders characteristically affecting the large muscles of the pelvic and shoulder girdles. Molecular genetic analyses have demonstrated causative mutations in the genes encoding a disparate collection of proteins involved in
Louise Glover et al.
Traffic (Copenhagen, Denmark), 8(7), 785-794 (2007-06-06)
The muscular dystrophies are a heterogeneous group of inherited disorders, defined by progressive muscle weakness and atrophy. Following the discovery of dystrophin, remarkable progress has been made in defining the molecular properties of proteins involved in the various dystrophies. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service