Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200385

Sigma-Aldrich

Monoclonal Anti-LMAN2L/VIPL antibody produced in mouse

clone VIPL-12, tissue culture supernatant

Synonym(s):

Monoclonal Anti-VIP36-like protein, Monoclonal Anti-lectin, Monoclonal Anti-mannose-binding 2-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

VIPL-12, monoclonal

form

buffered aqueous solution

mol wt

antigen ~34 kDa

species reactivity

human, rat, mouse

technique(s)

western blot: 1:100-1:200 using HEK-293T cells over-expressing human LMAN2L/VIPL.

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Lectin mannose binding 2-like (LMAN2L), also known as vesicular integral-membrane protein 36(VIP36) like protein (VIPL), is encoded by the gene mapped to human chromosome 2q11.2. It is a high-mannose type I membrane glycoprotein and is a member of the L-type lectin family. LMAN2L is expressed in wide variety of tissues.
Monoclonal Anti-LMAN2L/VIPL (mouse IgM isotype) is derived from the hybridoma VIPL-12 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide.

Specificity

Monoclonal Anti-LMAN2L/VIPL recognizes human LMAN2L/VIPL.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal of human LMAN2L, conjugated to KLH. The corresponding sequence is identical in mouse, rat, monkey, bovine and canine LMAN2L.

Application

Monoclonal Anti-LMAN2L/VIPL antibody produced in mouse may be used in immunoblotting.

Biochem/physiol Actions

Lectin mannose binding 2-like (LMAN2L) is a cargo receptor in the endoplasmic reticulum which has a role in glycoprotein transport. It may be implicated in targeting folded glycoproteins to the Golgi and misfolded glycoproteins to the ubiquitin-proteasome pathway. LMAN2L expression has been associated with bipolar disorder.
Lectin mannose-binding 2-like (LMAN2L) contains a luminal carbohydrate recognition domain (CRD) that binds to high-mannose type sugars in a Ca2+-dependent manner.

Physical form

Solution containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up toone month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation beforeuse. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profile-based data base scanning for animal L-type lectins and characterization of VIPL, a novel VIP36-like endoplasmic reticulum protein.
Nufer O
The Journal of Biological Chemistry, 278(18), 15886-15896 (2003)
Homozygous missense mutation in the LMAN2L gene segregates with intellectual disability in a large consanguineous Pakistani family.
Rafiullah R
Journal of medical Genetics, 53(2), 138-144 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service