Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200381

Sigma-Aldrich

Anti-LAYN (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Layilin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extracts of human HepG2 or HEK-293T cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAYN(143903)

General description

Layilin (LAYN) is a cell surface hyaluronan receptor, initially recognized as a binding partner of talin found in membrane ruffles of spreading cells. This transmembrane protein is broadly expressed in different cell types and tissue extracts. The protein LAYN belongs to the hyaluronan (HA)-binding protein family. The gene LAYN is mapped to the human chromosome location 11q23.1.

Specificity

Anti-LAYN (C-terminal) recognizes human LAYN.

Immunogen

synthetic peptide corresponding to the C-terminal region of human LAYN, conjugated to KLH. The antibody is affinity-purified using the immunizing peptide immobilized on agarose.

Application

Anti-LAYN (C-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Layilin (LAYN) binds to hyaluronan, a ubiquitous extracellular matrix component involved in the invasion of a variety of tumor cells, through its extracellular domain. The protein is implicated in cell adhesion and motility by interacting with the cytoskeletal-membrane linker proteins including merlin and radixin, through its cytoplasmic domain. The LAYN gene is overexpressed in human lung carcinoma A549. LAYN down-regulation by RNA interference inhibits invasion and lymphatic metastasis of A549 cells. The protein LAYN transfers signals from the extracellular matrix (ECM) to the cell cytoskeleton.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage,freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discardedif not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Layilin, a cell surface hyaluronan receptor, interacts with merlin and radixin
Bono P, et al.
Experimental Cell Research, 308(1), 177 -1187 (2005)
Pirin: a potential novel therapeutic target for castration-resistant prostate cancer regulated by miR-455-5p
Arai T, et al.
Molecular Oncology, 13(2), 322 -3337 (2019)
Down-regulation of layilin, a novel hyaluronan receptor, via RNA interference, inhibits invasion and lymphatic metastasis of human lung A549 cells
Chen Z, et al.
Biotechnology and Applied Biochemistry, 50(2), 89 -896 (2008)
Layilin, a novel integral membrane protein, is a hyaluronan receptor
Bono P, et al.
Molecular Biology of the Cell, 12(4), 891 -8900 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service