Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200377

Sigma-Aldrich

Anti-SMAD1 (Internal region) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-BSP1, Anti-JV4-1, Anti-JV41, Anti-MADH1, Anti-MADR1, Anti-SMAD family member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1-2 μg using lysates of HEK-293T cells over-expressing human SMAD1.
indirect immunofluorescence: 2.5-5.0 μg/mL using rat NRK cells.
western blot: 1-2 μg/mL using whole extracts of mouse F9 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD1(4086)
mouse ... Smad1(17125)
rat ... Smad1(25671)

General description

Mothers against decapentaplegic homolog 1 (SMAD1) belongs to receptor-activated Smads (R-SMADs) subfamily. It comprises Mad homology 1 (MH1) and nuclear localisation signal (NLS) in the N-terminus. The C-terminal region has the Mad homology 1 (MH2). SMAD1 also possesses leucine-rich nuclear export signal (NES) and a proline-tyrosine (PY) motif. SMAD1 gene is mapped to human chromosome 4q31.21.

Specificity

Anti-SMAD1 (internal region) recognizes human, mouse and rat SMAD1.

Immunogen

peptide corresponding to an internal region of human SMAD1, conjugated to KLH. The corresponding sequence is identical in mouse and differs by one amino acid in rat.

Application

Anti-SMAD1 (Internal region) antibody produced in rat may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Mothers against decapentaplegic homolog (SMADs), in general, mediate transmission of signals from the transforming growth factor-β (TGFβ) to the nucleus, and thus regulate multiple cellular processes, such as cell proliferation, apoptosis, and differentiation. SMAD1 is involved in mediating the signals of the bone morphogenetic proteins (BMP) and undergoes activation post phosphorylation by BMP receptor kinase. The phosphorylated form of SMAD1 forms a complex with SMAD4, which is important for its function in transcription regulation. SMAD1 is a target for SMAD-specific E3 ubiquitin ligases, such as Smad ubiquitin regulatory factor 1 (SMURF1 and SMURF2) and undergoes ubiquitination and proteasome-mediated degradation. SMAD1 is an oncogene, which favors cancer cell growth and invasion, especially in glioma. It is a potential biomarker in diabetic nephropathy (DN) pathology along with urinary IgG4. An elevated extracellular signal-regulated kinases (ERKs) lead to enhanced activation SMAD1 signaling, which is observed in the pathogenesis of Cardio‐facio‐cutaneous (CFC) syndrome. Alternatively spliced transcript variants encoding the same protein have been observed. Variants of SMAD1 and other SMAD genes are implicated in pulmonary arterial hypertension (PAH).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of the association between SMAD1 and HHIP gene variation and non-syndromic cleft-lip palate in Chilean case-parent trios
Suazo J, et al.
Genetics and Molecular Biology, 639-642 (2008)
Toshio Doi et al.
Diabetes, 67(5), 986-993 (2018-03-02)
Diabetic nephropathy (DN) is the major cause of end-stage kidney disease, but early biomarkers of DN risk are limited. Herein we examine urinary IgG4 and Smad1 as additional early DN biomarkers. We recruited 815 patients with type 2 diabetes; 554
Zhenguo Zeng et al.
Medicine, 99(10), e19451-e19451 (2020-03-10)
Non-small cell lung cancer (NSCLC) is the major cause of cancer mortality worldwide. Though multidisciplinary therapies have been widely used for NSCLC, its overall prognosis remains very poor, presumably owing to lack of effective prognostic biomarkers. SMAD, a well-known transcription
A Moustakas et al.
Journal of cell science, 114(Pt 24), 4359-4369 (2002-01-17)
Smad proteins transduce signals from transforming growth factor-beta (TGF-beta) superfamily ligands that regulate cell proliferation, differentiation and death through activation of receptor serine/threonine kinases. Phosphorylation of receptor-activated Smads (R-Smads) leads to formation of complexes with the common mediator Smad (Co-Smad)
Md Talat Nasim et al.
Human mutation, 32(12), 1385-1389 (2011-09-08)
Heterozygous germline mutations of BMPR2 contribute to familial clustering of pulmonary arterial hypertension (PAH). To further explore the genetic basis of PAH in isolated cases, we undertook a candidate gene analysis to identify potentially deleterious variation. Members of the bone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service