Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200371

Sigma-Aldrich

Anti-MAPKAP1 / SIN1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-MEKK2-interacting protein 1, Anti-MIP1, Anti-Mitogen-activated protein kinase associated protein 1, Anti-SAPK-Interacting protein 1, Anti-SIN1b, Anti-SIN1g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of mouse C2C12 cells.
western blot: 0.5-1.0 μg/mL using whole extracts of HEK-293T cells over-expressing human MAPKAP1.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Mitogen-activated protein kinase associated protein 1 (MAPKAP1 )/stress-activated protein kinase-interacting protein 1 (SIN1) is an essential subunit of the mammalian target of rapamycin complex 2 (mTORC2) complex.

Immunogen

synthetic peptide corresponding to an internal region within human MAPKAP1, conjugated to KLH. The corresponding sequence is identical in mouse, rat, canine and bovine.

Application

Anti-MAPKAP1/SIN1 antibody is suitable for use in immunoprecipitation (5-10 μg using lysates of mouse C2C12 cells) and western blot (0.5-1.0 μg/mL using whole extracts of HEK-293T cells over-expressing human MAPKAP1).

Biochem/physiol Actions

Mitogen-activated protein kinase associated protein 1 (MAPKAP1 )/stress-activated protein kinase-interacting protein 1 (SIN1) regulates cell survival and cytoskeletal organization. It maintains mammalian target of rapamycin complex 2 (mTORC2) integrity and modulates Akt phosphorylation. Knockdown of SIN1 results in the disruption of the rictor-mTOR interaction and in defective Akt-Ser473 phosphorylation.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Yang et al.
Genes & development, 20(20), 2820-2832 (2006-10-18)
Target of rapamycin (TOR) is an evolutionally conserved protein kinase in eukaryotes and a central cell growth controller. TOR exists in two distinct complexes, termed TORC1 and TORC2. Mammalian TORC2 has recently been shown to possess kinase activity toward the
Estela Jacinto et al.
Cell, 127(1), 125-137 (2006-09-12)
Mammalian target of rapamycin (mTOR) controls cell growth and proliferation via the raptor-mTOR (TORC1) and rictor-mTOR (TORC2) protein complexes. Recent biochemical studies suggested that TORC2 is the elusive PDK2 for Akt/PKB Ser473 phosphorylation in the hydrophobic motif. Phosphorylation at Ser473

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service