Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200368

Sigma-Aldrich

Anti-LRIG3 antibody, Mouse monoclonal

clone LRG3-3, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-LRIG3 antibody produced in mouse, Anti-FLJ90440, Anti-KIAA3016, Anti-leucine-rich repeats and immunoglobulin-like domains 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

LRG3-3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~125 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

flow cytometry: suitable
indirect immunofluorescence: suitable
western blot: 1.0-2.0 μg/mL using extracts of H1395 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRIG3(121227)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-LRIG3 (mouse IgG1 isotype) is derived from the hybridoma LRG3-3 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide corresponding to a sequence at the N-terminus human LRIG3 conjugated to KLH. Leucine-rich and immunoglobulin-like domain 3 (LRIG3) belongs to the family of LRIG. LRIG3 mRNA is widely expressed, with the highest levels in the stomach, skin and thyroid.

Immunogen

a peptide corresponding to a sequence at the N-terminus human LRIG3, conjugated to KLH. The corresponding sequence is identical in monkey and share high homology with rat and mouse LRIG3.

Application

Monoclonal Anti-LRIG3 antibody produced in mouse has been used in:
  • immunoblotting
  • immunofluorescence
  • flow cytometry (FACS)

Biochem/physiol Actions

Leucine-rich and immunoglobulin-like domain 3 (LRIG3) is implicated in tumorigenesis, psoriasis, neural crest development and complex tissue morphogenesis. LRIG3 interacts with the ErbB family of receptors and Netrin 1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The LRIG gene family has three vertebrate paralogs widely expressed in human and mouse tissues and a homolog in Ascidiacea
Guo D, et al.
Genomics, 84(1), 157-165 (2004)
Victoria E Abraira et al.
PloS one, 5(2), e8981-e8981 (2010-02-04)
The Lrig genes encode a family of transmembrane proteins that have been implicated in tumorigenesis, psoriasis, neural crest development, and complex tissue morphogenesis. Whether these diverse phenotypes reflect a single underlying cellular mechanism is not known. However, Lrig proteins contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service