Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200364

Sigma-Aldrich

Anti-PIWIL1 antibody, Mouse monoclonal

clone PIWIL1-6, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-PIWIL1 antibody produced in mouse, Anti-HIWI, Anti-MIWI, Anti-PIWI, Anti-piwi-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PIWIL1-6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~95 kDa

species reactivity

mouse, human

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded mouse testis and biotin / ExtrAvidin®-Peroxidase staining system
western blot: 2.5-5.0 μg/mL using whole extracts of mouse testis or HEK-293T cells over-expressing PIWIL1

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIWIL1(9271)
mouse ... Piwil1(57749)

General description

Monoclonal Anti-PIWIL1 (mouse IgG1 isotype) is derived from the hybridoma PIWIL1-6 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to an internal region of human PIWIL1 conjugated to keyhole limpet hemocyanin (KLH). The PIWI-like protein 1 (PIWIL1) belongs to the Argonaute protein family. This gene is located on human chromosome 12q24.33. PIWIL1 protein has 861 amino acid residues and possess a molecular weight of 98.6?kDa. It is expressed in prostate, ovary, brain, liver, heart, kidney and skeletal muscle.

Immunogen

synthetic peptide corresponding to an internal region of human PIWIL1, conjugated to KLH. The corresponding sequence is identical in mouse PIWIL1.

Application

Monoclonal Anti-PIWIL1 antibody produced in mouse has been used in various immunochemical techniques including immunoblotting (~95 kDa) and immunohistochemistry.

Biochem/physiol Actions

Piwi proteins and piRNAs have been implicated in epigenetic control of gene expression, transposon silencing and translation regulation. Piwi proteins participates in germline development and gametogenesis of many metazoan species. PIWI-like protein 1 (PIWIL1) is also involved in stem cell proliferation, embryogenesis, growth and development, along with differentiation and maturation in numerous organisms. It can act as a molecular marker for forecasting the prognosis of colorectal cancer (CRC) patients.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression status of PIWIL1 as a prognostic marker of colorectal cancer
Sun R, et al.
Disease Markers, 2017 (2017)
The biogenesis and function of PIWI proteins and piRNAs: progress and prospect
Thomson T and Lin H
Annual Review of Cell and Developmental, 25, 355-376 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service