Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200342

Sigma-Aldrich

Anti-SMAD3 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-HSPC193, Anti-HsT17436, Anti-JV15-2, Anti-MADH3, SMAD family member 3, Anti-mothers against decapentaplegic homolog 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using whole extracts of HEK-293T cells over-expressing human SMAD3.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD3(4088)
mouse ... Smad3(17127)
rat ... Smad3(25631)

General description

SMAD family member 3 (SMAD3) belongs to the receptor-regulated SMADs (R-SMADs) family. It is encoded by the gene mapped to human chromosome 15q22.33. SMAD3 is expressed in various tissues and is characterized with two conserved MH1 domains at N-terminal end and a MH2 domain at C-terminal end.

Specificity

Anti-SMAD3 recognizes human SMAD3.

Immunogen

peptide corresponding to an internal region of human SMAD3, conjugated to KLH. The corresponding sequence is identical in mouse, rat, monkey, dog, pig and bovine.

Biochem/physiol Actions

SMAD family member 3 (SMAD3) functions as an intracellular signaling molecule in the transforming growth factor β (TGF-β) pathway. Phosphorylated SMAD3 can form a complex with SMAD4, which translocates to the nucleus and regulates gene expression. SMAD3-SMAD4 complex can also interact with c-Jun and c-Fos at the activator protein 1 (AP-1) /SMAD site to regulate TGF-β-mediated transcription. SMAD3 has an inhibitory effect on wound healing. It also serves as a regulator of chondrogenesis and osteogenesis and inhibits early healing of bone fractures. SMAD3 gene polymorphisms leads to the development of osteoarthritis (OA).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of Smad3 gives rise to poor soft callus formation and accelerates early fracture healing
Kawakatsu M
Experimental and Molecular Pathology, 90(1), 107-115 (2011)
Crystal structure of a phosphorylated Smad2: Recognition of phosphoserine by the MH2 domain and insights on Smad function in TGF-$\beta$ signaling
Wu JW, et al.
Molecular Cell, 8(6), 1277-1289 (2001)
Atopic asthma after rhinovirus-induced wheezing is associated with DNA methylation change in the SMAD 3 gene promoter
Lund RJ, et al.
Allergy, 73(8), 1735-1740 (2018)
The polymorphism of SMAD3 rs1065080 is associated with increased risk for knee osteoarthritis
Lu C, et al.
Molecular Biology Reports, 46(4), 4501-4505 (2019)
The smads
Attisano L and Lee-Hoeflich ST
Genome Biology, 2(8), reviews3010-reviews3011 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service