SAB4200326
Anti-LRP1B (C-terminal region) antibody produced in rabbit
~1.5 mg/mL, affinity isolated antibody
Synonym(s):
Anti-LRP-DIT, Anti-LRPDIT, Anti-Low density lipoprotein receptor-related protein 1B
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~190 kDa (Rat)
antigen ~200 kDa (Human)
species reactivity
human, rat, mouse
concentration
~1.5 mg/mL
technique(s)
immunoprecipitation (IP): 5-10 μg using extracts of HEK-293T cells overexpressing human LRP1B.
indirect immunofluorescence: 1-2 μg/mL using HEK-293T cells overexpressing human LRP1B.
western blot: 1.5-3.0 μg/mL using extracts of rat brain (S1 fraction) and mouse brain (S1 fraction).
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... LRP1B(53353)
mouse ... Lrp1b(94217)
rat ... Lrp1b(311926)
General description
Immunogen
Application
- immunoblotting
- immunoprecipitation
- immunofluorescence
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service